Merck
  • New Derivatives of Natural Acyclic Guanidine Alkaloids with TRPV Receptor-Regulating Properties.
New Derivatives of Natural Acyclic Guanidine Alkaloids with TRPV Receptor-Regulating Properties.
Natural product communications (2015-09-29)
Ekaterina K Ogurtsova, Tatyana N Makarieva, Yuliya V Korolkova, Yaroslav A Andreev, Irina V Mosharova, Vladimir A Denisenko, Pavel S Dmitrenok, Yeon-Ju Lee, Eugene V Grishin, Valentin A Stonik
ZUSAMMENFASSUNG
The guanidine alkaloids, dihydropulchranin A (2), prepared from pulchranin A from the sponge Monanchora pulchra, and hexadecylguanidine (3), a synthetic analog of pulchranins, were studied for their TRPV channel-regulating activities. Compound 2 was active as an inhibitor of rTRPV1 and hTRPV3 receptors with EC50 values of 24.3 and 59.1 μM, respectively. Hexadecylguanidine (3) was not active against these receptors.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Hexadecylamin, 98%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
1H-Pyrazol-1-carboxamidin -hydrochlorid, 99%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Hexadecylamin, technical grade, 90%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.