Merck
Oncolytic viral therapy for neuroblastoma cells with Sindbis virus AR339 strain.
Pediatric surgery international (2015-08-25)
Ayako Takenouchi, Kengo Saito, Eriko Saito, Takeshi Saito, Tomoro Hishiki, Tadashi Matsunaga, Naohisa Isegawa, Hideo Yoshida, Naomi Ohnuma, Hiroshi Shirasawa
ZUSAMMENFASSUNG
With current treatment regimens, high-risk neuroblastoma (NB) remains largely incurable. Oncolytic viral therapy uses replication-competent viruses, like Sindbis virus (SINV), to kill cancers. The SINV AR339 strain is blood borne and relatively non-virulent. We evaluated the feasibility of SINV AR339 for treating human NB. The cytotoxicity and viral growth of SINV AR339 were evaluated for five human NB cell lines, SK-N-SH, IMR-32, LAN-5, GOTO, and RT-BM-1. SINV-induced apoptosis was confirmed by TUNEL assays and PARP-1 cleavage. In vivo effects of SINV on neuroblastoma cell xenografts in nude mice were assessed by intratumoral or intravenous SINV inoculation. In five human NB cell lines, SINV infections induced remarkable cytotoxicity. The mRNA expressions of anti-apoptotic genes, Bcl-2 and Bcl-xL, in LAN-5 and RT-BM-1, which were less sensitive to SINV infection, increased in response to SINV infection, while the other NB cell lines sensitive to SINV infection failed to respond. In nude mice, intratumoral and intravenous SINV inoculations caused significant regression of NB xenograft tumors. Our results suggested that SINV AR339 was significantly oncolytic against human NB. Thus, SINV showed promise as a novel therapy for treating NB.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Trizma® Base, Primary Standard and Buffer, ≥99.9% (titration), crystalline
Sigma-Aldrich
Trizma® Base, BioPerformance Certified, meets EP, USP testing specifications, suitable for cell culture, ≥99.9% (titration)
Sigma-Aldrich
Magnesiumchlorid -Lösung, Molecular Biology, 1.00 M±0.01 M
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Magnesiumchlorid, ≥98%
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Phenylmethansulfonylfluorid, ≥98.5% (GC)
Sigma-Aldrich
Tris(hydroxymethyl)aminomethan, ACS reagent, ≥99.8%
Sigma-Aldrich
Tromethamin, meets USP testing specifications
Sigma-Aldrich
Sigma 7-9®, ≥99% (titration), powder or crystals
Sigma-Aldrich
Trizma® Base, BioUltra, Molecular Biology, ≥99.8% (T)
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M
SAFC
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Magnesiumchlorid, powder, <200 μm
Sigma-Aldrich
Phenylmethansulfonylfluorid, ≥99.0% (T)
Sigma-Aldrich
Natriumchlorid, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Trizma® Base, ≥99.0% (T)
Sigma-Aldrich
Magnesiumchlorid -Lösung, BioUltra, Molecular Biology, 2 M in H2O
Sigma-Aldrich
Natriumchlorid, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Natriumchlorid -Lösung, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Natriumchlorid, 99.999% trace metals basis
Sigma-Aldrich
Magnesiumchlorid -Lösung, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Trizma® Base, BioXtra, pH 10.5-12.0 (1 M in H2O), ≥99.9% (titration)
Sigma-Aldrich
Trizma® Base, ≥99.9% (titration), crystalline
Sigma-Aldrich
Magnesiumchlorid, suitable for insect cell culture, BioReagent, ≥97.0%
Sigma-Aldrich
Trizma® Base, anhydrous, free-flowing, Redi-Dri, ≥99.9%
Sigma-Aldrich
Magnesiumchlorid -Lösung, PCR Reagent, 25 mM MgCI2 solution for PCR