Merck
Transcriptional regulator PRDM12 is essential for human pain perception.
Nature genetics (2015-05-26)
Ya-Chun Chen, Michaela Auer-Grumbach, Shinya Matsukawa, Manuela Zitzelsberger, Andreas C Themistocleous, Tim M Strom, Chrysanthi Samara, Adrian W Moore, Lily Ting-Yin Cho, Gareth T Young, Caecilia Weiss, Maria Schabhüttl, Rolf Stucka, Annina B Schmid, Yesim Parman, Luitgard Graul-Neumann, Wolfram Heinritz, Eberhard Passarge, Rosemarie M Watson, Jens Michael Hertz, Ute Moog, Manuela Baumgartner, Enza Maria Valente, Diego Pereira, Carlos M Restrepo, Istvan Katona, Marina Dusl, Claudia Stendel, Thomas Wieland, Fay Stafford, Frank Reimann, Katja von Au, Christian Finke, Patrick J Willems, Michael S Nahorski, Samiha S Shaikh, Ofélia P Carvalho, Adeline K Nicholas, Gulshan Karbani, Maeve A McAleer, Maria Roberta Cilio, John C McHugh, Sinead M Murphy, Alan D Irvine, Uffe Birk Jensen, Reinhard Windhager, Joachim Weis, Carsten Bergmann, Bernd Rautenstrauss, Jonathan Baets, Peter De Jonghe, Mary M Reilly, Regina Kropatsch, Ingo Kurth, Roman Chrast, Tatsuo Michiue, David L H Bennett, C Geoffrey Woods, Jan Senderek
ZUSAMMENFASSUNG
Pain perception has evolved as a warning mechanism to alert organisms to tissue damage and dangerous environments. In humans, however, undesirable, excessive or chronic pain is a common and major societal burden for which available medical treatments are currently suboptimal. New therapeutic options have recently been derived from studies of individuals with congenital insensitivity to pain (CIP). Here we identified 10 different homozygous mutations in PRDM12 (encoding PRDI-BF1 and RIZ homology domain-containing protein 12) in subjects with CIP from 11 families. Prdm proteins are a family of epigenetic regulators that control neural specification and neurogenesis. We determined that Prdm12 is expressed in nociceptors and their progenitors and participates in the development of sensory neurons in Xenopus embryos. Moreover, CIP-associated mutants abrogate the histone-modifying potential associated with wild-type Prdm12. Prdm12 emerges as a key factor in the orchestration of sensory neurogenesis and may hold promise as a target for new pain therapeutics.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonaler ANTI-FLAG® M2-Antikörper in Maus hergestellte Antikörper, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
L-Glutamin, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
Ethylendiamintetraessigsäure, 99.4-100.6%, powder
Sigma-Aldrich
L-Glutamin, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Ethylendiamintetraessigsäure, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylendiamintetraessigsäure -Lösung, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
SAFC
L-Glutamin
Sigma-Aldrich
Monoklonaler Anti-α-Tubulin-Antikörper in Maus hergestellte Antikörper, clone DM1A, ascites fluid
Sigma-Aldrich
Ethylendiamintetraessigsäure, 99.995% trace metals basis
Sigma-Aldrich
Ethylendiamintetraessigsäure, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Nitrotetrazolium-Blauchlorid, ≥90.0% (HPLC)
Sigma-Aldrich
L-Glutamin, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Ethylendiamintetraessigsäure, purified grade, ≥98.5%, powder
Sigma-Aldrich
Nitrotetrazolium-Blauchlorid, powder, electrophoresis grade
Sigma-Aldrich
L-Glutamin, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamin
Sigma-Aldrich
Anti-Histon H3-Antikörper, CT, pan, serum, Upstate®
Sigma-Aldrich
Vasoaktives intestinales Peptid, Mensch, Schwein, Ratte, ≥95% (HPLC), powder
Sigma-Aldrich
Stickstoff-14N2, 99.99 atom % 14N
Sigma-Aldrich
Vasoaktives intestinales Peptid, Mensch, Schwein, Ratte, synthetic, ≥95% (HPLC), powder