Merck
  • Transformation with oncogenic Ras and the simian virus 40 T antigens induces caspase-dependent sensitivity to fatty acid biosynthetic inhibition.
Transformation with oncogenic Ras and the simian virus 40 T antigens induces caspase-dependent sensitivity to fatty acid biosynthetic inhibition.
Journal of virology (2015-04-10)
Shihao Xu, Cody M Spencer, Joshua Munger
ZUSAMMENFASSUNG
Oncogenesis is frequently accompanied by the activation of specific metabolic pathways. One such pathway is fatty acid biosynthesis, whose induction is observed upon transformation of a wide variety of cell types. Here, we explored how defined oncogenic alleles, specifically the simian virus 40 (SV40) T antigens and oncogenic Ras(12V), affect fatty acid metabolism. Our results indicate that SV40/Ras(12V)-mediated transformation of fibroblasts induces fatty acid biosynthesis in the absence of significant changes in the concentration of fatty acid biosynthetic enzymes. This oncogene-induced activation of fatty acid biosynthesis was found to be mammalian target of rapamycin (mTOR) dependent, as it was attenuated by rapamycin treatment. Furthermore, SV40/Ras(12V)-mediated transformation induced sensitivity to treatment with fatty acid biosynthetic inhibitors. Pharmaceutical inhibition of acetyl-coenzyme A (CoA) carboxylase (ACC), a key fatty acid biosynthetic enzyme, induced caspase-dependent cell death in oncogene-transduced cells. In contrast, isogenic nontransformed cells were resistant to fatty acid biosynthetic inhibition. This oncogene-induced sensitivity to fatty acid biosynthetic inhibition was independent of the cells' growth rates and could be attenuated by supplementing the medium with unsaturated fatty acids. Both the activation of fatty acid biosynthesis and the sensitivity to fatty acid biosynthetic inhibition could be conveyed to nontransformed breast epithelial cells through transduction with oncogenic Ras(12V). Similar to what was observed in the transformed fibroblasts, the Ras(12V)-induced sensitivity to fatty acid biosynthetic inhibition was independent of the proliferative status and could be attenuated by supplementing the medium with unsaturated fatty acids. Combined, our results indicate that specific oncogenic alleles can directly confer sensitivity to inhibitors of fatty acid biosynthesis. Viral oncoproteins and cellular mutations drive the transformation of normal cells to the cancerous state. These oncogenic alterations induce metabolic changes and dependencies that can be targeted to kill cancerous cells. Here, we find that the cellular transformation resulting from combined expression of the SV40 early region with an oncogenic Ras allele is sufficient to induce cellular susceptibility to fatty acid biosynthetic inhibition. Inhibition of fatty acid biosynthesis in these cells resulted in programmed cell death, which could be rescued by supplementing the medium with nonsaturated fatty acids. Similar results were observed with the expression of oncogenic Ras in nontransformed breast epithelial cells. Combined, our results suggest that specific oncogenic alleles induce metabolic dependencies that can be exploited to selectively kill cancerous cells.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Trypanblau -Lösung, 0.4%, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Rapamycin, ≥95% (HPLC), powder
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Trypanblau, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Rapamycin, Ready Made Solution, 2.5 mg/mL in DMSO (2.74 mM), from Streptomyces hygroscopicus
Sigma-Aldrich
Dimethylsulfoxid, PCR Reagent
Sigma-Aldrich
Trypanblau, ≥80% (HPLC), Dye content 60 %
Sigma-Aldrich
DL-Glyceraldehyd-3-phosphat -Lösung, 45-55 mg/mL in H2O
Sigma-Aldrich
TOFA, ≥98% (HPLC)
Sigma-Aldrich
Dimethylsulfoxid, ≥99.6%, ReagentPlus®