Merck
  • Structural insights into methanol-stable variants of lipase T6 from Geobacillus stearothermophilus.
Structural insights into methanol-stable variants of lipase T6 from Geobacillus stearothermophilus.
Applied microbiology and biotechnology (2015-06-01)
Adi Dror, Margarita Kanteev, Irit Kagan, Shalev Gihaz, Anat Shahar, Ayelet Fishman
ZUSAMMENFASSUNG
Enzymatic production of biodiesel by transesterification of triglycerides and alcohol, catalyzed by lipases, offers an environmentally friendly and efficient alternative to the chemically catalyzed process while using low-grade feedstocks. Methanol is utilized frequently as the alcohol in the reaction due to its reactivity and low cost. However, one of the major drawbacks of the enzymatic system is the presence of high methanol concentrations which leads to methanol-induced unfolding and inactivation of the biocatalyst. Therefore, a methanol-stable lipase is of great interest for the biodiesel industry. In this study, protein engineering was applied to substitute charged surface residues with hydrophobic ones to enhance the stability in methanol of a lipase from Geobacillus stearothermophilus T6. We identified a methanol-stable variant, R374W, and combined it with a variant found previously, H86Y/A269T. The triple mutant, H86Y/A269T/R374W, had a half-life value at 70 % methanol of 324 min which reflects an 87-fold enhanced stability compared to the wild type together with elevated thermostability in buffer and in 50 % methanol. This variant also exhibited an improved biodiesel yield from waste chicken oil compared to commercial Lipolase 100L® and Novozyme® CALB. Crystal structures of the wild type and the methanol-stable variants provided insights regarding structure-stability correlations. The most prominent features were the extensive formation of new hydrogen bonds between surface residues directly or mediated by structural water molecules and the stabilization of Zn and Ca binding sites. Mutation sites were also characterized by lower B-factor values calculated from the X-ray structures indicating improved rigidity.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Schwefelsäure, ACS reagent, 95.0-98.0%
Sigma-Aldrich
Ethylacetat, ACS reagent, ≥99.5%
Sigma-Aldrich
Diethylether, anhydrous, ACS reagent, ≥99.0%, contains BHT as inhibitor
Sigma-Aldrich
Acetonitril, ACS reagent, ≥99.5%
Sigma-Aldrich
Diethylether, ACS reagent, anhydrous, ≥99.0%, contains BHT as inhibitor
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Schwefelsäure, 99.999%
Sigma-Aldrich
Oleinsäure, technical grade, 90%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Palmitinsäure, ≥99%
Sigma-Aldrich
Zirkonium(IV)-oxychlorid Octahydrat, reagent grade, 98%
Sigma-Aldrich
α-Linoleic acid, ≥98%
Sigma-Aldrich
Linolensäure, ≥99%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Oleinsäure, ≥99% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
4-Nitrophenol, ReagentPlus®, ≥99%
Sigma-Aldrich
Ethylacetat, anhydrous, 99.8%
Sigma-Aldrich
Stearinsäure, Grade I, ≥98.5% (capillary GC)
Sigma-Aldrich
Diethylether, contains 1 ppm BHT as inhibitor, anhydrous, ≥99.7%
Sigma-Aldrich
Oleinsäure, suitable for cell culture, BioReagent
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
L-Lysin -monohydrochlorid, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
Linolsäure, liquid, BioReagent, suitable for cell culture
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Heptadecansäure, ≥98%
Sigma-Aldrich
Stearinsäure, reagent grade, 95%
Sigma-Aldrich
Palmitinsäure, BioXtra, ≥99%