Merck
  • Statin-mediated inhibition of cholesterol synthesis induces cytoprotective autophagy in human leukemic cells.
Statin-mediated inhibition of cholesterol synthesis induces cytoprotective autophagy in human leukemic cells.
European journal of pharmacology (2015-09-12)
Urosh Vilimanovich, Mihajlo Bosnjak, Andrija Bogdanovic, Ivanka Markovic, Aleksandra Isakovic, Tamara Kravic-Stevovic, Aleksandar Mircic, Vladimir Trajkovic, Vladimir Bumbasirevic
ZUSAMMENFASSUNG
Statins exhibit anti-leukemic properties due to suppression of the mevalonate pathway by the inhibition of 3-hydroxy-3-methylglutaryl coenzyme A reductase, and subsequent depletion of cholesterol, farnesylpyrophosphate, and geranylgeranylpyrophosphate. We investigated the role of autophagy, a controlled intracellular self-digestion, in the anti-leukemic action of statins. Treatment with low concentrations (≤6 µM) of statins, cholesterol depletion, and specific inhibition of cholesterol synthesis and protein farnesylation or geranylgeranylation, all inhibited proliferation of leukemic cell lines and primary leukemic cells without inducing overt cell death. Statins and agents that selectively reduce intracellular cholesterol levels, but not the inhibition of protein farnesylation or geranylgeranylation, induced autophagy in leukemic cells. The observed autophagic response was associated with the reduction of phosphorylated Akt levels in the lipid rafts, accompanied by a decrease in the activation of the main autophagy suppressor mammalian target of rapamycin (mTOR) and its substrate ribosomal p70S6 kinase (p70S6K). No significant autophagy induction and downregulation of mTOR/p70S6K activation were observed in normal leukocytes. Autophagy suppression by bafilomycin A1 or RNA interference-mediated knockdown of beclin-1 and microtubule-associated protein 1 light chain 3B induced apoptotic death in statin-treated leukemic cells, an effect attenuated by the addition of mevalonate or squalene, but not farnesylpyrophosphate or geranylgeranylpyrophosphate. Therefore, while the inhibition of cholesterol synthesis, protein farnesylation, and geranylgeranylation all contributed to anti-leukemic effects of statins, the inhibition of cholesterol synthesis was solely responsible for the induction of cytoprotective autophagy. These data indicate that combined treatment with statins and autophagy inhibitors might be potentially useful in anti-leukemic therapy.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Cholesterin, Sigma Grade, ≥99%
Sigma-Aldrich
Glutaraldehyd -Lösung, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Sigma-Aldrich
Osmiumtetroxid, ReagentPlus®, 99.8%
Sigma-Aldrich
Glutaraldehyd -Lösung, Grade II, 25% in H2O
Sigma-Aldrich
Glutaraldehyd -Lösung, 50 wt. % in H2O
Sigma-Aldrich
Cholesterin, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Osmiumtetroxid -Lösung, 4 wt. % in H2O
Sigma-Aldrich
Fluoreszein-5(6)-isothiocyanat, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
Squalen, ≥98%, liquid
Sigma-Aldrich
SyntheChol® NS0-Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Sigma-Aldrich
Osmiumtetroxid -Lösung, 2.5 wt. % in tert-butanol
Sigma-Aldrich
Rapamycin, Ready Made Solution, 2.5 mg/mL in DMSO (2.74 mM), from Streptomyces hygroscopicus
Sigma-Aldrich
Cholesterin, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Glutaraldehyd -Lösung, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Osmiumtetroxid, Sealed ampule.
Sigma-Aldrich
1-Hexen, ≥99%
Sigma-Aldrich
Osmiumtetroxid -Lösung, suitable for electron microscopy, 4% in H2O
Supelco
Cholesterin -Lösung, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
1-Hexen, 97%
Sigma-Aldrich
Osmiumtetroxid, ACS reagent, ≥98.0%
Sigma-Aldrich
4-Methylvaleriansäure, 99%
Supelco
Squalen, analytical standard
Sigma-Aldrich
Glutaraldehyd -Lösung, Grade I, 70% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
1-Hexen, 97%
Sigma-Aldrich
Fluoreszein-5(6)-isothiocyanat, ≥90% (HPLC)
Sigma-Aldrich
Fluorescein-Isothiocyanat Isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Osmiumtetroxid -Lösung, suitable for electron microscopy, 2% in H2O
Sigma-Aldrich
Glutarsäure-Dialdehyd -Lösung, 50 wt. % in H2O, FCC
Sigma-Aldrich
Glutaraldehyd -Lösung, Grade I, 8% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyd -Lösung, 50% in H2O, suitable for photographic applications