- Fluorescence enhancement of coumarin-6-sulfonyl chloride amino acid derivatives in cyclodextrin media.
Fluorescence enhancement of coumarin-6-sulfonyl chloride amino acid derivatives in cyclodextrin media.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry (2002-05-07)
S M Al-Kindy, F E Suliman, A A Al-Hamadi
PMID11990574
ZUSAMMENFASSUNG
Coumarin-6-sulfonyl (6-CS) amino acid derivatives form inclusion complexes with a- and /-cyclodextrins (CD) in aqueous solution. The stoichiometry of the inclusion complex and the equilibrium constant were investigated. Using a fluorescence technique and alanine-beta-CD as a model, a 1:2 guest-host complex was established, and K = 4.7 x 10(5) mol(-2) l(2) was obtained. Fluorescence enhancement was observed for all derivatives studied, with glycine exhibiting a greater enhancement, and tyrosine showing the least. The stability of the inclusion complex was found to depend on the respective sizes of the guest-host complex and their interaction.