Merck
  • Human induced pluripotent stem cells differentiate into insulin-producing cells able to engraft in vivo.
Human induced pluripotent stem cells differentiate into insulin-producing cells able to engraft in vivo.
Acta diabetologica (2015-03-04)
Silvia Pellegrini, Federica Ungaro, Alessia Mercalli, Raffaella Melzi, Guido Sebastiani, Francesco Dotta, Vania Broccoli, Lorenzo Piemonti, Valeria Sordi
ZUSAMMENFASSUNG
New sources of insulin-secreting cells are strongly required for the cure of diabetes. Recent successes in differentiating embryonic stem cells, in combination with the discovery that it is possible to derive human induced pluripotent stem cells (iPSCs) from somatic cells, have raised the possibility that patient-specific beta cells might be derived from patients through cell reprogramming and differentiation. In this study, we aimed to obtain insulin-producing cells from human iPSCs and test their ability to secrete insulin in vivo. Human iPSCs, derived from both fetal and adult fibroblasts, were differentiated in vitro into pancreas-committed cells and then transplanted into immunodeficient mice at two different stages of differentiation (posterior foregut and endocrine cells). IPSCs were shown to differentiate in insulin-producing cells in vitro, following the stages of pancreatic organogenesis. At the end of the differentiation, the production of INSULIN mRNA was highly increased and 5 ± 2.9 % of the cell population became insulin-positive. Terminally differentiated cells also produced C-peptide in vitro in both basal and stimulated conditions. In vivo, mice transplanted with pancreatic cells secreted human C-peptide in response to glucose stimulus, but transplanted cells were observed to lose insulin secretion capacity during the time. At histological evaluation, the grafts resulted to be composed of a mixed population of cells containing mature pancreatic cells, but also pluripotent and some neuronal cells. These data overall suggest that human iPSCs have the potential to generate insulin-producing cells and that these differentiated cells can engraft and secrete insulin in vivo.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
D-(+)-Glukose, ≥99.5% (GC)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
D-(+)-Glukose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
3-Isobutyl-1-methylxanthin, ≥99% (HPLC), powder
Sigma-Aldrich
Magnesiumchlorid -Lösung, Molecular Biology, 1.00 M±0.01 M
Sigma-Aldrich
Formaldehyd -Lösung, Molecular Biology, 36.5-38% in H2O
Sigma-Aldrich
Magnesiumchlorid, ≥98%
Sigma-Aldrich
Formaldehyd -Lösung, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
SAFC
Formaldehyd -Lösung, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Sigma-Aldrich
Dextrose, 97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications
Sigma-Aldrich
3-Isobutyl-1-methylxanthin, BioUltra, ≥99%
Sigma-Aldrich
D-(+)-Glukose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Magnesiumchlorid, powder, <200 μm
Sigma-Aldrich
Formaldehyd -Lösung, Molecular Biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
HEPES-Pufferlösung, 1 M in H2O
Sigma-Aldrich
D-(+)-Glukose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-(+)-Glukose, ACS reagent
Supelco
D-(+)-Glukose, analytical standard
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
Magnesiumchlorid -Lösung, BioUltra, Molecular Biology, 2 M in H2O
Sigma-Aldrich
Magnesiumchlorid -Lösung, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Formaldehyd -Lösung, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Aphidicolin, ≥98% (HPLC), powder
Supelco
Formaldehyd -Lösung, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
Magnesiumchlorid, suitable for insect cell culture, BioReagent, ≥97.0%
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
Magnesiumchlorid -Lösung, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
D-(+)-Glukose, suitable for mouse embryo cell culture, ≥99.5% (GC)