Merck
  • Subtype-specific analysis of the K65R substitution in HIV-1 that confers hypersusceptibility to a novel nucleotide-competing reverse transcriptase inhibitor.
Subtype-specific analysis of the K65R substitution in HIV-1 that confers hypersusceptibility to a novel nucleotide-competing reverse transcriptase inhibitor.
Antimicrobial agents and chemotherapy (2015-03-18)
Hong-Tao Xu, Susan P Colby-Germinario, Peter K Quashie, Richard Bethell, Mark A Wainberg
ZUSAMMENFASSUNG
Compound A is a novel nucleotide-competing HIV-1 reverse transcriptase (RT) inhibitor (NcRTI) that selects for a unique W153L substitution that confers hypersusceptibility to tenofovir, while the K65R substitution in RT confers resistance against tenofovir and enhances susceptibility to NcRTIs. Although the K65R substitution is more common in subtype C viruses, the impact of subtype variability on NcRTI susceptibility has not been studied. In the present study, we performed experiments with compound A by using purified recombinant RT enzymes and viruses of subtypes B and C and circulating recombinant form CRF_A/G. We confirmed the hypersusceptibility of K65R substitution-containing RTs to compound A for subtype C, CRF_A/G, and subtype B. Steady-state kinetic analysis showed that K65R RTs enhanced the susceptibility to compound A by increasing binding of the inhibitor to the nucleotide binding site of RT in a subtype-independent manner, without significantly discriminating against the natural nucleotide substrate. These data highlight the potential utility of NcRTIs, such as compound A, for treatment of infections with K65R substitution-containing viruses, regardless of HIV-1 subtype.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Glycerin, Molecular Biology, ≥99.0%
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Harnstoff, powder, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
L-Glutamin, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Harnstoff, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Glycerin, ≥99.5%
Sigma-Aldrich
IPTG, ≥99% (TLC), ≤0.1% Dioxane
Sigma-Aldrich
L-Glutamin, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Glycerin -Lösung, 83.5-89.5% (T)
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M
Supelco
Harnstoff, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
Glycerin, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
SAFC
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Glycerin, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Harnstoff -Lösung, BioUltra, ~8 M in H2O
SAFC
L-Glutamin
Sigma-Aldrich
Glycerin, BioXtra, ≥99% (GC)
Sigma-Aldrich
Harnstoff, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Harnstoff, BioUltra, Molecular Biology, 99% (T)
Sigma-Aldrich
Natriumchlorid, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Harnstoff, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Natriumchlorid, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Natriumchlorid -Lösung, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Natriumchlorid, 99.999% trace metals basis
Sigma-Aldrich
Glycerin, FCC, FG
Sigma-Aldrich
Glycerin, meets USP testing specifications