Merck
Vasodilatory Effect of Wogonin on the Rat Aorta and Its Mechanism Study.
Biological & pharmaceutical bulletin (2015-12-04)
Jing-Tian Qu, Dong-Xuan Zhang, Fang Liu, Hao-Ping Mao, Ya-Ke Ma, Yue Yang, Chun-Xiao Li, Li-Zhen Qiu, Xiao Geng, Jian-Mei Zhang, Xiu-Mei Gao, Lu Chen, Hong Wang
ZUSAMMENFASSUNG
Wogonin, a natural flavonoid, is one of the bioactive compounds of the medicinal herb Eucommia ulmoides OLIV. widely used in southeastern Asia for treating hypertension. However, the molecular mechanisms for the therapeutic benefits remain largely unclear. The present study investigated the vasodilatory effect of wogonin and its possible mechanisms. The flavonoid (0.1-100 µM) caused concentration-dependent relaxations in endothelium-intact aortic rings precontracted with norepinephrine (NE, 1 µM) or potassium chloride (KCl, 60 mM). Preincubation with wogonin (10, 100 µM) for 20 min significantly inhibited the contractile responses to NE (0.1, 1, 10 µM) or KCl (7.5, 15, 30, 60 mM). Relaxant responses to wogonin were not inhibited by N(G)-nitro-L-arginine methylester (100 µM) or endothelial denudation. In a Ca(2+)-free Krebs' solution, wogonin not only blocked Ca(2+) influx-dependent vasoconstriction by either NE (1 µM) or KCl (100 mM), but also inhibited NE (1 µM)-induced tonic contraction, which is dependent on intracellular Ca(2+) release. Wogonin also suppressed the elevation of [Ca(2+)]i induced by KCl (60 mM) after exhausting the calcium store in sarcoplasmic and endoplasmic reticula with thapsigargin (1 µM) or by ATP (100 µM) in primary vascular smooth muscle cells. These findings suggest that wogonin-induced responses are mainly due to the inhibition of both intracellular Ca(2+) release and extracellular Ca(2+) influx.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Kaliumphosphat, ACS reagent, ≥99.0%
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Kaliumchlorid, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Natriumbicarbonat, ACS reagent, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Magnesiumsulfat, anhydrous, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Natriumbicarbonat, powder, BioReagent, Molecular Biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Natriumbicarbonat, ReagentPlus®, ≥99.5%, powder
Sigma-Aldrich
Kaliumphosphat, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Kaliumchlorid, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Kaliumchlorid, Molecular Biology, ≥99.0%
Sigma-Aldrich
Kaliumchlorid, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Kaliumchlorid, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E508, 99-100.5% (AT), ≤0.0001% Al
Sigma-Aldrich
Ethylenglykol-bis(2-aminoethylether)-N,N,N′,N′-Tetraessigsäure, Molecular Biology, ≥97.0%
Sigma-Aldrich
Magnesiumsulfat, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Kaliumchlorid, puriss. p.a., ≥99.5% (AT)
Sigma-Aldrich
Natriumbicarbonat, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.7%
Sigma-Aldrich
Natriumbicarbonat, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E500, 99.0-100.5%, powder
Sigma-Aldrich
Kaliumphosphat, ReagentPlus®
Sigma-Aldrich
Kaliumchlorid -Lösung, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Magnesiumsulfat, puriss. p.a., drying agent, anhydrous, ≥98.0% (KT), powder (very fine)
Sigma-Aldrich
Kaliumphosphat, Molecular Biology, ≥98.0%
Sigma-Aldrich
Kaliumchlorid, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Magnesiumsulfat, BioReagent, suitable for cell culture, suitable for insect cell culture