Merck
  • Visible light absorption and photoelectrochemical activity of colorless molecular 1,3-bis(dicyanomethylidene)indane (BDMI) by surface complexation on TiO₂.
Visible light absorption and photoelectrochemical activity of colorless molecular 1,3-bis(dicyanomethylidene)indane (BDMI) by surface complexation on TiO₂.
Physical chemistry chemical physics : PCCP (2015-06-26)
Soo-Byung Ko, Tae In Ryu, An-Na Cho, Junichi Fujisawa, Hiroshi Segawa, Nam-Gyu Park
ZUSAMMENFASSUNG
Adsorption of the colorless 1,3-bis(dicyanomethylidene)indane (BDMI) onto a nanocrystalline TiO2 surface unusually turned the BDMI a deep blue color. Upon contact of the BDMI-adsorbed TiO2 (BDMI-TiO2) with an iodide-based redox electrolyte, a photocurrent density as high as 14.9 mA cm(-2) was generated with a photovoltage of 0.42 V, leading to a power conversion efficiency of 3.63%. This unprecedented photovoltaic performance was simultaneously investigated by spectroscopic studies of BDMI-TiO2 films and density functional theory (DFT)/time-dependent DFT (TD-DFT) computational approaches for [BDMI](-)[Ti(OH)3·H2O](+) (1) as a simple model compound to inspect the light to current conversion abilities. All these results established that the color change from colorless to deep blue and the highly efficient photocurrent generation through binding on the TiO2 surface originates from interfacial charge transfer transitions from anionic BDMI to TiO2.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Titan(IV)isopropoxid, 97%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Laurinsäure, ≥98%, FCC, FG
Sigma-Aldrich
Titan(IV)isopropoxid, 99.999% trace metals basis
Sigma-Aldrich
1-Butanol, anhydrous, 99.8%
Sigma-Aldrich
Titan(IV)isopropoxid, ≥97.0%
Sigma-Aldrich
Essigsäure, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
5α-Androstan-17β-ol-3-on, ≥97.5%
Sigma-Aldrich
1-Butanol, Molecular Biology, ≥99%
Sigma-Aldrich
Laurinsäure, 98%
Sigma-Aldrich
Terpineol, mixture of isomers
Sigma-Aldrich
Essigsäure, ≥99.5%, FCC, FG
Sigma-Aldrich
Essigsäure, natural, ≥99.5%, FG
Sigma-Aldrich
Butylalkohol, natural, ≥99.5%, FCC, FG
Sigma-Aldrich
Laurinsäure, natural, ≥98%, FCC, FG
Sigma-Aldrich
Dodecansäure, ≥99% (GC/titration)
Sigma-Aldrich
Titan(IV)isopropoxid, packaged for use in deposition systems
Sigma-Aldrich
5α-Androstan-17β-ol-3-on, purum, ≥99.0% (TLC)
Sigma-Aldrich
Essigsäure-12C2, 99.9 atom % 12C