Merck
  • Identification of XRCC1 Arg399Gln and XRCC3 Thr241Met Polymorphisms in a Turkish Population and Their Association with the Risk of Chronic Lymphocytic Leukemia.
Identification of XRCC1 Arg399Gln and XRCC3 Thr241Met Polymorphisms in a Turkish Population and Their Association with the Risk of Chronic Lymphocytic Leukemia.
Indian journal of hematology & blood transfusion : an official journal of Indian Society of Hematology and Blood Transfusion (2015-06-19)
Pelin Mutlu, Mualla Pınar Elçi, Murat Yıldırım, Oral Nevruz, Ahmet Türker Çetin, Ferit Avcu
ZUSAMMENFASSUNG
DNA repair systems are essential for cellular functions. Defects due to sequence variations in DNA repair genes can lead severe failure of cell functions and causing many cancer types including leukemia. The aim of this study was to investigate the relationship between XRCC1 Arg399Gln and XRCC3 Thr241Met polymorphisms and susceptibility to chronic lymphocytic leukemia (CLL) in Turkish patients. In addition, genotype distribution of these polymorphisms was compared with other populations. The frequencies of Arg399Gln and Thr241Met single nucleotide polymorphisms were studied in 25 CLL patients and 30 healthy individuals. Single nucleotide polymorphisms were genotyped by PCR-RFLP method. The genotype and allele frequencies of Arg399Gln and Thr241Met polymorphisms were not statistically different between the CLL patients and control group. The allelic frequency similarities were found between Turkish and Brazilian populations for Arg399Gln polymorphism. On the other hand, similarities were found between Turkish and other Caucasian populations for Thr241Met polymorphism. Marked differences were observed between American African versus Turkish and Chinese versus Turkish populations for Arg399Gln and Thr241Met polymorphisms respectively. These results indicate that Arg399Gln and Thr241Met polymorphisms were not associated with the development of CLL in Turkish population and ethnic differences is one of the most important factor for allele frequency differences.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Magnesiumchlorid -Lösung, Molecular Biology, 1.00 M±0.01 M
Sigma-Aldrich
Magnesiumchlorid, ≥98%
Sigma-Aldrich
Ethidiumbromid -Lösung, BioReagent, Molecular Biology, 10 mg/mL in H2O
Sigma-Aldrich
Magnesiumchlorid, powder, <200 μm
Sigma-Aldrich
Magnesiumchlorid -Lösung, BioUltra, Molecular Biology, 2 M in H2O
Sigma-Aldrich
Magnesiumchlorid -Lösung, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Ethidiumbromid -Lösung, BioReagent, Molecular Biology, 500 μg/mL in H2O
Sigma-Aldrich
Ethidiumbromid, BioReagent, Molecular Biology, powder
Sigma-Aldrich
Magnesiumchlorid, suitable for insect cell culture, BioReagent, ≥97.0%
Sigma-Aldrich
Magnesiumchlorid -Lösung, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Ethidiumbromid, ~95% (HPLC)
Sigma-Aldrich
Magnesiumchlorid, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Ethidiumbromid -Lösung, suitable for fluorescence, ~1% in H2O
Sigma-Aldrich
Magnesiumchlorid, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Supelco
Pyridaben, PESTANAL®, analytical standard
Sigma-Aldrich
Magnesiumchlorid -Lösung, 0.1 M
Sigma-Aldrich
Magnesiumchlorid -Lösung, BioUltra, Molecular Biology, ~0.025 M in H2O