Merck
  • Tailoring material properties of cardiac matrix hydrogels to induce endothelial differentiation of human mesenchymal stem cells.
Tailoring material properties of cardiac matrix hydrogels to induce endothelial differentiation of human mesenchymal stem cells.
ACS applied materials & interfaces (2015-05-07)
Megan E Jeffords, Jinglei Wu, Mickey Shah, Yi Hong, Ge Zhang
ZUSAMMENFASSUNG
Cardiac matrix hydrogel has shown great promise as an injectable biomaterial due to the possession of cardiac-specific extracellular matrix composition. A cardiac matrix hydrogel facilitating neovascularization will further improve its therapeutic outcomes in cardiac repair. In this study, we explored the feasibility of tailoring material properties of cardiac matrix hydrogels using a natural compound, genipin, to promote endothelial differentiation of stem cells. Our results demonstrated that the genipin cross-linking could increase the mechanical properties of the cardiac matrix hydrogel to a stiffness range promoting endothelial differentiation of human mesenchymal stem cells (hMSCs). It also decreased the swelling ratio and prolonged degradation without altering gelation time. Human mesenchymal stem cells cultured on the genipin cross-linked cardiac matrix hydrogels showed great viability. After 1 day culture, hMSCs demonstrated down-regulation of early endothelial marker expression and up-regulation of mature endothelial marker expression. Especially for 1 mM genipin cross-linked cardiac matrix hydrogels, hMSCs showed particularly significant expression of mature endothelial cell marker vWF. These attractive results indicate the potential of using genipin cross-linked cardiac matrix hydrogels to promote rapid vascularization for cardiac infarction treatment through minimally invasive therapy.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumdodecylsulfat, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Glycin, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Glycin, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Formaldehyd -Lösung, Molecular Biology, 36.5-38% in H2O
Sigma-Aldrich
Natriumdodecylsulfat, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Salzsäure -Lösung, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Formaldehyd -Lösung, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Calciumchlorid -Lösung, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Natriumdodecylsulfat, ACS reagent, ≥99.0%
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Natriumdodecylsulfat, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Phenol -Lösung, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, BioReagent, Molecular Biology
Sigma-Aldrich
Calciumchlorid, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Glycin, BioUltra, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Glycin, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Salzsäure, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
Phenol, puriss., ≥99.5% (GC), meets analytical specification of Ph. Eur., BP, USP, crystalline (detached)
Sigma-Aldrich
Natriumdodecylsulfat, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, 20% in H2O
Supelco
Salzsäure -Lösung, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Formaldehyd -Lösung, Molecular Biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Supelco
Natriumdodecylsulfat, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Phenol -Lösung, Saturated with 0.01 M citrate buffer, pH 4.3 ± 0.2, BioReagent, Molecular Biology
Sigma-Aldrich
Phenol, ≥99%
Sigma-Aldrich
Isoamylalkohol, ≥98%, FG
Sigma-Aldrich
Verflüssigtes Phenol, ≥89.0%