Merck
  • Synthesis of Polyamidoamine Dendrimer for Encapsulating Tetramethylscutellarein for Potential Bioactivity Enhancement.
Synthesis of Polyamidoamine Dendrimer for Encapsulating Tetramethylscutellarein for Potential Bioactivity Enhancement.
International journal of molecular sciences (2015-11-12)
Daniel M Shadrack, Egid B Mubofu, Stephen S Nyandoro
ZUSAMMENFASSUNG
The biomedical potential of flavonoids is normally restricted by their low water solubility. However, little has been reported on their encapsulation into polyamidoamine (PAMAM) dendrimers to improve their biomedical applications. Generation four (G4) PAMAM dendrimer containing ethylenediaminetetraacetic acid core with acrylic acid and ethylenediamine as repeating units was synthesized by divergent approach and used to encapsulate a flavonoid tetramethylscutellarein (TMScu, 1) to study its solubility and in vitro release for potential bioactivity enhancement. The as-synthesized dendrimer and the dendrimer-TMScu complex were characterized by spectroscopic and spectrometric techniques. The encapsulation of 1 into dendrimer was achieved by a co-precipitation method with the encapsulation efficiency of 77.8% ± 0.69% and a loading capacity of 6.2% ± 0.06%. A phase solubility diagram indicated an increased water solubility of 1 as a function of dendrimer concentration at pH 4.0 and 7.2. In vitro release of 1 from its dendrimer complex indicated high percentage release at pH 4.0. The stability study of the TMScu-dendrimer at 0, 27 and 40 °C showed the formulations to be stable when stored in cool and dark conditions compared to those stored in light and warmer temperatures. Overall, PAMAM dendrimer-G4 is capable of encapsulating 1, increasing its solubility and thus could enhance its bioactivity.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ethylacetat, ACS reagent, ≥99.5%
Sigma-Aldrich
Tetrahydrofuran, contains 200-400 ppm BHT as inhibitor, ACS reagent, ≥99.0%
Sigma-Aldrich
Acetonitril, ACS reagent, ≥99.5%
Sigma-Aldrich
Tetrahydrofuran, anhydrous, ≥99.9%, inhibitor-free
Sigma-Aldrich
Tetrahydrofuran, anhydrous, contains 250 ppm BHT as inhibitor, ≥99.9%
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
N,N′-Dicyclohexylcarbodiimid, 99%
Sigma-Aldrich
Ethylacetat, anhydrous, 99.8%
Sigma-Aldrich
Acetonitril, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitril, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitril, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Ethylacetat, ≥99%, FCC, FG
Sigma-Aldrich
N,N′-Dicyclohexylcarbodiimid -Lösung, 1.0 M in methylene chloride
Sigma-Aldrich
Ethylacetat, ACS reagent, ≥99.5%
Supelco
Tetrahydrofuran, HPLC grade, ≥99.9%, inhibitor-free
Sigma-Aldrich
Ethylacetat
Sigma-Aldrich
Acetonitril, ReagentPlus®, 99%
Sigma-Aldrich
Ethylacetat, natural, ≥99%, FCC, FG
Sigma-Aldrich
Ethylacetat, biotech. grade, ≥99.8%
Sigma-Aldrich
Ethylacetat, ReagentPlus®, ≥99.8%
Sigma-Aldrich
trans-2-[3-(4-tert-Butylphenyl)-2-methyl-2-propenyliden]malononitril, ≥98%