Merck
Liposomal delivery of lipoarabinomannan triggers Mycobacterium tuberculosis specific T-cells.
Tuberculosis (Edinburgh, Scotland) (2015-06-06)
Stephanie Kallert, Sebastian F Zenk, Paul Walther, Mark Grieshober, Tanja Weil, Steffen Stenger
ZUSAMMENFASSUNG
Lipoarabinomannan (LAM) is a major cell wall component of Mycobacterium tuberculosis (Mtb). LAM specific human T-lymphocytes release interferon-γ (IFNγ) and have antimicrobial activity against intracellular Mtb suggesting that they contribute to protection. Therefore the induction of LAM-specific memory T-cells is an attractive approach for the design of a new vaccine against tuberculosis. A prerequisite for the activation of LAM-specific T-cells is the efficient uptake and transport of the glycolipid antigen to the CD1 antigen presenting machinery. Based on the hydrophobicity of LAM we hypothesized that packaging of LAM into liposomes will support the activation of T-lymphocytes. We prepared liposomes containing phosphatidylcholine, cholesterol, stearylated octaarginine and LAM via thin layer hydration method (LIPLAM). Flow cytometry analysis using fluorescently labelled LIPLAM showed an efficient uptake by antigen presenting cells. LAM delivered via liposomes was biologically active as demonstrated by the down-regulation of peroxisome proliferator activated receptor gamma (PPARγ) protein expression. Importantly, LIPLAM induced higher IFNγ production by primary human T-lymphocytes than purified LAM (2-16 times) or empty liposomes. These results suggest that the delivery of mycobacterial glycolipids via liposomes is a promising approach to promote the induction of M. tuberculosis specific T-cell responses.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Glycerin, Molecular Biology, ≥99.0%
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Natriumdodecylsulfat, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Glycin, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
2-Mercaptoethanol, Molecular Biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sigma-Aldrich
Glycin, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Cholesterin, Sigma Grade, ≥99%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Natriumdodecylsulfat, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Natriumdodecylsulfat, ACS reagent, ≥99.0%
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Propidiumjodid, ≥94.0% (HPLC)
Sigma-Aldrich
Deoxycholsäure Natriumsalz, BioXtra, ≥98.0% (dry matter, NT)
Sigma-Aldrich
Deoxycholsäure Natriumsalz, ≥97% (titration)
Sigma-Aldrich
Natriumdodecylsulfat, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Glycerin, ≥99.5%
Sigma-Aldrich
Glycerin -Lösung, 83.5-89.5% (T)
Sigma-Aldrich
2-Mercaptoethanol, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Glycin, BioUltra, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Glycin, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Natriumdodecylsulfat, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Cholesterin, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Glycerin, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, 20% in H2O
Sigma-Aldrich
Glycerin, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Glycerin, BioXtra, ≥99% (GC)
Sigma-Aldrich
HEPES-Pufferlösung, 1 M in H2O
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer