Merck
  • Upregulation of Multidrug Resistance-Associated Protein 1 by Allyl Isothiocyanate in Human Bronchial Epithelial Cell: Involvement of c-Jun N-Terminal Kinase Signaling Pathway.
Upregulation of Multidrug Resistance-Associated Protein 1 by Allyl Isothiocyanate in Human Bronchial Epithelial Cell: Involvement of c-Jun N-Terminal Kinase Signaling Pathway.
Oxidative medicine and cellular longevity (2015-08-15)
Shujun Wang, Shanshan Wang, Chenyin Wang, Yajun Chen, Jie Li, Xueqi Wang, Dianlei Wang, Zegeng Li, Zhaoliang Peng, Ling Fan
ZUSAMMENFASSUNG
Multidrug resistance-associated protein 1 (MRP1) plays a protective role in the etiology and progression of chronic obstructive pulmonary disease (COPD) which results from oxidative stress and inflammation of lung injury. The lower functional MRP1 activity is related to COPD development. Our previous study showed that Allyl isothiocyanate (AITC) induced the expression and activity of MRP1 in a dose-dependent manner. However, which signaling pathway contributes to the upregulation of MRP1 by AITC is unclear. In this study, signaling pathway specific inhibitors were used to examine the mechanism of AITC. We found that JNK inhibitor SP600125 treatment decreased MRP1 mRNA expression in 16HBE14o- cells. But the ERK inhibitor U0126 or PI3K/Akt inhibitor LY294002 produced no obvious effect. The AITC-induced increase of MRP1 mRNA expression was abolished by cotreatment of SP600125, while it was not obviously affected by U0126 or LY294002. Furthermore, AITC acivates the JNK signaling pathway in 16HBE14o- cells. Finally, we found that JNK pathway mediated the upregulation of AITC-induced expression and function of MRP1. Taken together, our results indicated that AITC increased the expression and the activity of MRP1 via a JNK-dependent pathway. ERK and PI3K signaling pathway were not involved in the expression of MRP1 mRNA.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Natriumdodecylsulfat, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Natriumdodecylsulfat, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Natriumdodecylsulfat, ACS reagent, ≥99.0%
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Phenylmethansulfonylfluorid, ≥98.5% (GC)
Sigma-Aldrich
Natriumdodecylsulfat, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Natriumdodecylsulfat, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, 20% in H2O
Sigma-Aldrich
Phenylmethansulfonylfluorid, ≥99.0% (T)
Supelco
Natriumdodecylsulfat, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Natriumdodecylsulfat, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Dimethylsulfoxid, PCR Reagent
Sigma-Aldrich
Natriumdodecylsulfat, ≥98.0% (GC)
Sigma-Aldrich
Natriumdodecylsulfat, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
SP600125, ≥98% (HPLC)
Sigma-Aldrich
Natriumdodecylsulfat, ≥90% ((Assay))
Sigma-Aldrich
Natriumdodecylsulfat, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
2,2′-Bichinolin-4,4′-dicarbonsäure Dinatriumsalz Dihydrat, ≥98% (HPLC)
Sigma-Aldrich
5-Carboxyfluorescein, BioReagent, suitable for fluorescence, ≥95% (HPLC)
Sigma-Aldrich
8-Octanoyloxypyren-1,3,6-Trisulfonsäure Trinatriumsalz, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Natriumdodecylsulfat, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Dimethylsulfoxid, ≥99.6%, ReagentPlus®