Merck
Isolation and culture of human oligodendrocyte precursor cells from neurospheres.
Brain research bulletin (2015-09-01)
Yabin Lu, Yinxiang Yang, Zhaoyan Wang, Caiying Wang, Qingan Du, Qian Wang, Zuo Luan
ZUSAMMENFASSUNG
Culture of human oligodendrocyte precursor cells (OPCs) can help understand the regulatory mechanism of differentiation and myelination of oligodendrocytes. However, existing culture methods have limitations, particularly the lack of a source of human donor tissue and high cost. We sorted cells with the A2B5(+)PSA-NCAM(-) phenotype from neurospheres instead of human donor tissues through immunomagnetic sorting and subsequently cultured the isolated cells in OPC medium. Of all the isolated cells, 15.69% were of the A2B5(+)PSA-NCAM(-) phenotype. More than 90% of the isolated OPCs expressed the OPC-specific markers O4, PDGFαR, and Sox10, and less than 5% of cells expressed GFAP and Tuj-1. After induction, the isolated cells had the capacity to differentiate into oligodendrocytes. Furthermore, the OPCs could be stably passaged in vitro for at least four generations and all the cells had high expression levels of O4 and Sox10 and very low expression levels of GFAP and Tuj-1; moreover, the cells had the capacity to differentiate into oligodendrocytes. After four passages, OPCs can proliferate at least 14 times above. In addition, in the presence of B27, only one cytokine, namely, bFGF, was sufficient to maintain proliferation, and this greatly reduced the experimental cost. Cells of the A2B5(+)PSA-NCAM(-) phenotype have already been identified as OPCs. We developed and characterized a reproducible, simple, and economical method for the isolation and culture of human OPCs. This method will contribute to studying the function of OPCs in development, disease, and treatment.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Heparin Natriumsalz aus Schweinedarm, Grade I-A, ≥180 USP units/mg, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
L-Glutamin, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
hEGF, EGF, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
Bromwasserstoffsäure, ACS reagent, 48%
Sigma-Aldrich
Ethylendiamintetraessigsäure, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Bromwasserstoffsäure, reagent grade, 48%
Sigma-Aldrich
L-Glutamin, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Ethylendiamintetraessigsäure, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylendiamintetraessigsäure -Lösung, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrogenbromid -Lösung, 33 wt. % in acetic acid
SAFC
L-Glutamin
Sigma-Aldrich
Ethylendiamintetraessigsäure, 99.995% trace metals basis
Sigma-Aldrich
Ethylendiamintetraessigsäure, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
EGF human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
Bromwasserstoffsäure, 48 wt. % in H2O, ≥99.99%
Sigma-Aldrich
L-Glutamin, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Epidermaler Wachstumsfaktor human, EGF, recombinant, expressed in Escherichia coli, >97% (SDS-PAGE)
Sigma-Aldrich
Ethylendiamintetraessigsäure, purified grade, ≥98.5%, powder
Sigma-Aldrich
L-Glutamin, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamin