Merck
  • 4-Hydroxynonenal: A Superior Oxidative Biomarker Compared to Malondialdehyde and Carbonyl Content Induced by Carbon Tetrachloride in Rats.
4-Hydroxynonenal: A Superior Oxidative Biomarker Compared to Malondialdehyde and Carbonyl Content Induced by Carbon Tetrachloride in Rats.
Journal of toxicology and environmental health. Part A (2015-08-08)
Dong Hyun Kim, Seung Jun Kwack, Kyung Sik Yoon, Jin Shil Choi, Byung-Mu Lee
ZUSAMMENFASSUNG
Carbon tetrachloride (CCl4), a halogenated substance that generates free radical species during metabolism in vivo, induces hepatotoxicity, produces oxidative DNA damage, and increased levels of protein carbonyl, malondialdehyde (MDA), and 4-hydroxynonenal (4-HNE). In this study, Sprague-Dawley rats received single or repeated ip injections of carbon tetrachloride (CCl4), and formation and persistence of carbonyls, MDA, and 4-HNE in plasma were measured using gas chromatography-mass spectrometry. After a single injection of 500 mg/kg CCl4 the in vivo half-lives of MDA and carbonyl content were 1.5 d and 2 d, respectively, while that of 4-HNE was approximately 10 d. Treatment with CCl4 (50, 100, 500, or 1000 mg/kg) dose-dependently increased these oxidative biomarkers in blood. However, formation of protein carbonyls and MDA was less sensitive than 4-HNE to CCl4. Levels of serum glutamic oxaloacetic transaminase (SGOT) and glutamic pyruvic transaminase (SGPT) (hepatotoxicity markers) rose with CCl4 doses. After a single injection (500 mg/kg), the peak level of SGOT was observed after 8 h but SGPT after 24 h. Overall, 4-HNE was more dose-sensitive and showed greater formation subchronically than other biomarkers. Multiple ip treatments with 300 mg CCl4 /kg (d 1, 3, 6, 10, 14, and 21) demonstrated that 4-HNE formation was highest (18-fold, peak/control) and subchronic up to d 21 (last treatment day), unlike other biomarkers. Data suggest that 4-HNE, MDA, and carbonyl content may be useful oxidative biomarkers for exposure to free radical generating halogenated compounds. However, 4-HNE appears to be a more sensitive and sustainable biomarker for toxicological and risk assessments.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Kaliumchlorid, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Magnesiumsulfat, anhydrous, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Kaliumchlorid, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Kaliumchlorid, Molecular Biology, ≥99.0%
Sigma-Aldrich
Kaliumchlorid, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
2,6-Di-tert-butyl-4-Methylphenol, ≥99.0% (GC), powder
Sigma-Aldrich
Kaliumchlorid, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E508, 99-100.5% (AT), ≤0.0001% Al
Sigma-Aldrich
Magnesiumsulfat, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Kaliumchlorid, puriss. p.a., ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M
SAFC
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Butyliertes Hydroxytoluol, ≥99%, FCC, FG
Sigma-Aldrich
Kaliumchlorid -Lösung, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Magnesiumsulfat, puriss. p.a., drying agent, anhydrous, ≥98.0% (KT), powder (very fine)
Sigma-Aldrich
Kaliumchlorid, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Magnesiumsulfat, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Natriumchlorid, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Natriumchlorid -Lösung, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Natriumchlorid, 99.999% trace metals basis
Sigma-Aldrich
Kaliumchlorid, BioXtra, ≥99.0%
Sigma-Aldrich
Magnesiumsulfat, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥97%
Sigma-Aldrich
Magnesiumsulfat -Lösung, Molecular Biology, 1.00 M±0.04 M
Sigma-Aldrich
Kaliumchlorid, 99.999% trace metals basis
Sigma-Aldrich
Magnesiumsulfat -Lösung, BioUltra, Molecular Biology