Merck
Chocolate smells pink and stripy: Exploring olfactory-visual synesthesia.
Cognitive neuroscience (2015-04-22)
Alex Russell, Richard J Stevenson, Anina N Rich
ZUSAMMENFASSUNG
Odors are often difficult to identify, and can be perceived either via the nose or mouth ("flavor"; not usually perceived as a "smell"). These features provide a unique opportunity to contrast conceptual and perceptual accounts of synesthesia. We presented six olfactory-visual synesthetes with a range of odorants. They tried to identify each smell, evaluate its attributes and illustrate their elicited visual experience. Judges rated the similarity of each synesthetes' illustrations over time (test-retest reliability). Synesthetic images were most similar from the same odor named consistently, but even inconsistently named same odors generated more similar images than different odors. This was driven by hedonic similarity. Odors presented as flavors only resulted in similar images when consistently named. Thus, the primary factor in generating a reliable synesthetic image is the name, with some influence of odor hedonics. Hedonics are a basic form of semantic knowledge, making this consistent with a conceptual basis for synaesthetic links.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Saccharose, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Saccharose, ≥99.5% (GC)
Sigma-Aldrich
Saccharose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Saccharose, BioUltra, Molecular Biology, ≥99.5% (HPLC)
Sigma-Aldrich
Zitronensäure, ACS reagent, ≥99.5%
Sigma-Aldrich
Saccharose, ≥99.5% (GC)
Sigma-Aldrich
Zitronensäure, ≥99.5%, FCC, FG
Sigma-Aldrich
Saccharose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Zitronensäure, 99%
Sigma-Aldrich
Saccharose, ACS reagent
Sigma-Aldrich
Zitronensäure, anhydrous, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
1-Butanol, anhydrous, 99.8%
Sigma-Aldrich
Phenylacetylen, 98%
Sigma-Aldrich
Saccharose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sigma-Aldrich
Essigsäure, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Zitronensäure, BioUltra, anhydrous, ≥99.5% (T)
Sigma-Aldrich
5α-Androstan-17β-ol-3-on, ≥97.5%
Sigma-Aldrich
1-Butanol, Molecular Biology, ≥99%
Sigma-Aldrich
Saccharose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
cis-3-Hexen-1-ol, natural, >98%, FCC, FG
Sigma-Aldrich
Essigsäure, ≥99.5%, FCC, FG
Sigma-Aldrich
Zitronensäure, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
Sigma-Aldrich
Essigsäure, natural, ≥99.5%, FG
Sigma-Aldrich
Saccharose, ≥99% (GC), Grade I, suitable for plant cell culture
Sigma-Aldrich
Butylalkohol, natural, ≥99.5%, FCC, FG
Sigma-Aldrich
2-Hydroxybuttersäure, 97%
Sigma-Aldrich
Saccharose, meets USP testing specifications
Sigma-Aldrich
cis-3-Hexen-1-ol, 98%
Sigma-Aldrich
5α-Androstan-17β-ol-3-on, purum, ≥99.0% (TLC)
Sigma-Aldrich
Fenchol, ≥96%, FG