Merck
  • Enzymes enhance degradation of the fiber-starch-protein matrix of distillers dried grains with solubles as revealed by a porcine in vitro fermentation model and microscopy.
Enzymes enhance degradation of the fiber-starch-protein matrix of distillers dried grains with solubles as revealed by a porcine in vitro fermentation model and microscopy.
Journal of animal science (2015-05-29)
R Jha, T A Woyengo, J Li, M R Bedford, T Vasanthan, R T Zijlstra
ZUSAMMENFASSUNG
Effects of treating corn and wheat distillers dried grains with solubles (DDGS) with a multicarbohydrase alone or in combination with a protease on porcine in vitro fermentation characteristics and the matrix structure of the DGGS before and after the fermentation were studied. Three DDGS samples (wheat DDGS sample 1 [wDDGS1], wheat DDGS sample 2 [wDDGS2], and corn DDGS [cDDGS]) were predigested with pepsin and pancreatin. Residues were then subjected to in vitro fermentation using buffered mineral solution inoculated with fresh pig feces without or with a multicarbohydrase alone or in combination with protease in a 3 × 3 factorial arrangement. Accumulated gas production was measured for up to 72 h. Concentration of VFA was measured in fermented solutions. The matrix of native DDGS and their residues after fermentation was analyzed using confocal laser scanning microscopy and scanning electron microscopy to determine internal and external structures, respectively. On a DM basis, wDDGS1, wDDGS2, and cDDGS contained 35.5, 43.4, and 29.0% CP; 2.23, 0.51, and 6.40% starch; 0.82, 0.80, and 0.89% available Lys; and 24.8, 22.5, and 23.0% total nonstarch polysaccharides, respectively. The in vitro digestibility of DM for wDDGS1, wDDGS2, and cDDGS was 67.7, 72.1, and 59.6%, respectively. The cDDGS had greater ( < 0.05) total gas and VFA production than both wheat DDGS. The wDDGS2 had lower ( < 0.05) total gas production than wDDGS1. Multicarbohydrase increased ( < 0.05) total gas production for cDDGS and total VFA production for wDGGS1 but did not increase gas or VFA production for wDDGS2. Addition of protease with multicarbohydrase to DDGS reduced ( < 0.05) total gas and VFA productions and increased ( < 0.05) branched-chain VFA regardless of DDGS type. Confocal laser scanning microscopy and scanning electron microscopy revealed that DDGS were mainly aggregates of resistant and nonfermentable starchy and nonstarchy complexes formed during DDGS production. After in vitro fermentation with porcine fecal inoculum, particles of enzyme-treated DDGS were generally smaller than those of the untreated DDGS. In conclusion, cDDGS had a more porous matrix that was more fermentable than the wheat DDGS. The wDDGS2 was less fermentable than wDDGS1. Multicarbohydrase increased fermentability of cDDGS and wDDGS1 but not wDDGS2, indicating that its efficacy in DDGS is dependent on matrix porosity and DDGS source. Protease hindered efficacy of multicarbohydrase.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Wasser, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
Saccharose, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Wasser, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Saccharose, ≥99.5% (GC)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
(3-Aminopropyl)triethoxysilan, 99%
Sigma-Aldrich
Phosphorsäure, 85 wt. % in H2O, 99.99% trace metals basis
Sigma-Aldrich
Chloramphenicol, ≥98% (HPLC)
Sigma-Aldrich
Saccharose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Saccharose, BioUltra, Molecular Biology, ≥99.5% (HPLC)
Sigma-Aldrich
Natriumcyanoborhydrid, reagent grade, 95%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Wasser, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Wasser, Molecular Biology, sterile filtered
Sigma-Aldrich
Saccharose, ≥99.5% (GC)
Sigma-Aldrich
Pancreatin aus Schweinepankreas, 4 × USP specifications
Sigma-Aldrich
Saccharose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Phosphorsäure, crystalline, ≥99.999% trace metals basis
Sigma-Aldrich
Saccharose, ACS reagent
Sigma-Aldrich
(3-Aminopropyl)triethoxysilan, ≥98.0%
Sigma-Aldrich
Chloramphenicol, BioReagent, suitable for plant cell culture
Sigma-Aldrich
(3-Aminopropyl)triethoxysilan, ≥98%
Sigma-Aldrich
Phosphorsäure -Lösung, 85 wt. % in H2O, FCC, FG
Sigma-Aldrich
Wasser, BioPerformance Certified
Sigma-Aldrich
Saccharose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sigma-Aldrich
Essigsäure, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Phosphorsäure, BioUltra, ≥85% (T)
Sigma-Aldrich
5α-Androstan-17β-ol-3-on, ≥97.5%
Sigma-Aldrich
Saccharose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Essigsäure, ≥99.5%, FCC, FG