Merck
  • The analysis of the acute phase response during the course of Trypanosoma carassii infection in the goldfish (Carassius auratus L.).
The analysis of the acute phase response during the course of Trypanosoma carassii infection in the goldfish (Carassius auratus L.).
Developmental and comparative immunology (2015-06-28)
Nikolina Kovacevic, Mariel O Hagen, Jiasong Xie, Miodrag Belosevic
ZUSAMMENFASSUNG
The expression of genes encoding the acute phase proteins (APP) during the course of Trypanasoma carassii infection in the goldfish was determined using quantitative PCR. Significant changes in the mRNA levels of ceruloplasmin (Cp), C-reactive protein (CRP), transferrin (Tf), hemopexin (Hx) and serum amyloid A (SAA) were observed in the kidney, liver and spleen at various days post infection (dpi). Of the five acute phase protein genes examined, CRP and SAA exhibited the highest expression in the tissues during the acute infection. Cp and Tf were up-regulated throughout the acute course of infection in the liver. During the chronic phase of the infection, APP expression in the liver was similar to that in the non-infected control fish. At 7 dpi, Cp, Tf and Hx were down-regulated in the spleen, and Cp and Tf kidney, but their mRNA levels gradually returned to those of control non-infected fish. In contrast, during the chronic phase of the infection, there was an up-regulation of Cp, Hx and Tf in the spleen, and Tf and SAA in the kidney. The goldfish CRP was cloned and functionally characterized. CRP was differentially expressed in normal goldfish immune cells, with highest expression in monocytes and lowest expression in mature macrophages. A recombinant goldfish CRP (rgfCRP) was generated using prokaryotic expression. rgfCRP enhanced complement-mediated killing of trypanosomes in vitro, and the lysis increased after addition of immune serum. rgfCRP did not affect the production of reactive oxygen and nitrogen intermediates by monocytes and macrophages, respectively.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Phorbol-12-myristat-13-acetat, ≥99% (TLC), film or powder
Sigma-Aldrich
Imidazol, ReagentPlus®, 99%
Sigma-Aldrich
PMA, for use in molecular biology applications, ≥99% (HPLC), Molecular Biology
Sigma-Aldrich
Harnstoff, powder, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Imidazol, ACS reagent, ≥99% (titration)
Sigma-Aldrich
L-Glutathion reduziert, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
L-Glutathion reduziert, ≥98.0%
Sigma-Aldrich
Harnstoff, ACS reagent, 99.0-100.5%
Sigma-Aldrich
IPTG, ≥99% (TLC), ≤0.1% Dioxane
Sigma-Aldrich
Ethylendiamintetraessigsäure, 99.4-100.6%, powder
Sigma-Aldrich
Imidazol, Molecular Biology, ≥99% (titration)
Sigma-Aldrich
Ethylendiamintetraessigsäure, anhydrous, crystalline, BioReagent, suitable for cell culture
Supelco
Harnstoff, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
Ethylendiamintetraessigsäure -Lösung, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Harnstoff -Lösung, BioUltra, ~8 M in H2O
Sigma-Aldrich
Imidazol, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Ethylendiamintetraessigsäure, 99.995% trace metals basis
Sigma-Aldrich
Ethylendiamintetraessigsäure, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Imidazol-Pufferlösung, BioUltra, 1 M in H2O
Sigma-Aldrich
Harnstoff, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Harnstoff, BioUltra, Molecular Biology, 99% (T)
Sigma-Aldrich
Imidazol, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Harnstoff, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Nitrotetrazolium-Blauchlorid, ≥90.0% (HPLC)
Sigma-Aldrich
N-Chlorsuccinimid, 98%
Sigma-Aldrich
Phorbol-12-myristat-13-acetat, synthetic, ≥98.0% (TLC)
Sigma-Aldrich
Harnstoff, suitable for electrophoresis
Sigma-Aldrich
Isopropyl β-D-1-Thiogalactopyranosid, ≥99% (TLC)
Sigma-Aldrich
Isopropyl-β-D-thiogalactopyranosid -Lösung, ReadyMade IPTG solution for Blue-white screening
Sigma-Aldrich
Harnstoff, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)