Merck
  • Cortistatin is dysregulated in skin tissue of patients with psoriasis vulgaris and suppresses keratinocyte proliferation in vitro.
Cortistatin is dysregulated in skin tissue of patients with psoriasis vulgaris and suppresses keratinocyte proliferation in vitro.
International journal of dermatology (2015-06-23)
Dai Li, Hui-Lan He, Ming-Zhu Yao, Ming-Liang Chen, Xiang Chen
ZUSAMMENFASSUNG
Psoriasis is characterized by the unregulated proliferation of epidermal keratinocytes and increased expression of proinflammatory mediators in the skin. Cortistatin, an endogenous cyclic neuropeptide, inhibits the proliferation of inflammatory cells. We investigated the expression of cortistatin in patients with psoriasis vulgaris and examined its effects on keratinocyte growth in vitro. Serum levels of cortistatin were determined by enzyme-linked immunosorbent assay (ELISA) in 72 patients with psoriasis vulgaris and 76 age-matched healthy volunteers. Cortistatin expression was also examined by immunohistochemistry of skin biopsies from 14 patients and 14 healthy subjects. The effects of cortistatin on the proliferation of primary keratinocytes were assessed using the 3-(4,5-dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide (MTT) assay and BrdU incorporation assay. Intracellular levels of cAMP in keratinocytes in the presence or absence of cortistatin were determined by ELISA. Serum levels of cortistatin and expression levels in skin were significantly lower in patients with psoriasis than in healthy subjects. Cortistatin inhibited keratinocyte proliferation in vitro in a dose-dependent manner and substantially reduced intracellular cAMP levels in keratinocytes. Cortistatin is downregulated in the skin of patients with psoriasis vulgaris and suppresses keratinocyte growth in vitro.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
3-Isobutyl-1-methylxanthin, ≥99% (HPLC), powder
Sigma-Aldrich
(3-Aminopropyl)triethoxysilan, 99%
Sigma-Aldrich
Thiazolylblau-Tetrazoliumbromid, 98%
Sigma-Aldrich
hEGF, EGF, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
Thiazolylblau-Tetrazoliumbromid, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
3-Isobutyl-1-methylxanthin, BioUltra, ≥99%
Sigma-Aldrich
Perchlorsäure, 70%, 99.999% trace metals basis
Sigma-Aldrich
(3-Aminopropyl)triethoxysilan, ≥98.0%
Sigma-Aldrich
(3-Aminopropyl)triethoxysilan, ≥98%
Sigma-Aldrich
EGF human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
Zyklisches Adenosin-3′,5′-Monophosphat, ≥98.5% (HPLC), powder
Sigma-Aldrich
Epidermaler Wachstumsfaktor human, EGF, recombinant, expressed in Escherichia coli, >97% (SDS-PAGE)
Sigma-Aldrich
(3-Aminopropyl)triethoxysilan, packaged for use in deposition systems, ≥98%
Sigma-Aldrich
Adenosine 3′,5′-cyclic monophosphate tris salt, ≥97% (HPLC), powder
Sigma-Aldrich
MISSION® esiRNA, targeting human CENPS, APITD1-CORT