Merck
  • Co-delivery of paclitaxel and gemcitabine via CD44-targeting nanocarriers as a prodrug with synergistic antitumor activity against human biliary cancer.
Co-delivery of paclitaxel and gemcitabine via CD44-targeting nanocarriers as a prodrug with synergistic antitumor activity against human biliary cancer.
Biomaterials (2015-04-22)
Ilkoo Noh, Hyun-Ouk Kim, Jihye Choi, Yuna Choi, Dong Ki Lee, Yong-Min Huh, Seungjoo Haam
ZUSAMMENFASSUNG
Multi-drug delivery focuses on different signaling pathways in cancer cells that have synergistic anti-proliferative effects. In this study, we developed multi-prodrug nanocarriers (MPDNCs) consisting of poly (l-lysine)-carboxylate PTX (PLL-PTX) and hyaluronic acid-conjugated GEM (HA-GEM) for CD44-targeted synergistic biliary cancer therapy. An in vitro study of cell viability and mRNA expression levels and an in vivo study showed that MPDNCs more effectively inhibit proliferation in CD44-overexpressing cancer cells (HuCCT1) than in cells with lower CD44 expression (SCK) by synergistically inducing apoptosis. Consequently, these results demonstrate that MPDNCs are prodrugs with synergistic cancer therapeutic efficacy and effective cellular uptake at target cells compared to free drugs, indicating their strong potential as efficient multi-drug-carrying nano-platforms for cancer treatment.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Deuteriumoxid, 99.9 atom % D
Sigma-Aldrich
Dimethylsulfoxid-d6, 99.9 atom % D
Sigma-Aldrich
Chloroform-d, 99.8 atom % D
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
Dimethylsulfoxid-d6, 99.9 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Natriumazid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
N-Hydroxysuccinimid, 98%
Sigma-Aldrich
Dimethylsulfoxid-d6, 99.5 atom % D
Sigma-Aldrich
Deuteriumoxid, 99.9 atom % D, contains 0.05 wt. % 3-(trimethylsilyl)propionic-2,2,3,3-d4 acid, sodium salt
Sigma-Aldrich
Natriumazid, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Diethylether, contains 1 ppm BHT as inhibitor, anhydrous, ≥99.7%
Sigma-Aldrich
Fluoreszein-5(6)-isothiocyanat, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
HEPES-Pufferlösung, 1 M in H2O
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.05 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 1 % (v/v) TMS
Sigma-Aldrich
Gemcitabin-Hydrochlorid -hydrochlorid, ≥98% (HPLC)
Sigma-Aldrich
Natriumazid, purum p.a., ≥99.0% (T)
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
Natriumazid, BioXtra
Sigma-Aldrich
Diethylether
Sigma-Aldrich
Dimethylsulfoxid-d6, "100%", 99.96 atom % D
Sigma-Aldrich
Deuteriumoxid, 99.9 atom % D, contains 0.75 wt. % 3-(trimethylsilyl)propionic-2,2,3,3-d4 acid, sodium salt
Sigma-Aldrich
Acetonitril, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.1 % (v/v) TMS
Sigma-Aldrich
Dimethylsulfoxid-d6, 99.9 atom % D, contains 1 % (v/v) TMS
Sigma-Aldrich
Dimethylsulfoxid-d6, anhydrous, 99.9 atom % D