Merck
  • Investigating detrusor muscle concentrations of oxybutynin after intravesical delivery in an ex vivo porcine model.
Investigating detrusor muscle concentrations of oxybutynin after intravesical delivery in an ex vivo porcine model.
Journal of pharmaceutical sciences (2015-05-20)
Nicholas A Williams, Kay M Lee, Chris J Allender, Jenna L Bowen, Mark Gumbleton, Tim Harrah, Aditya Raja, Hrishi B Joshi
ZUSAMMENFASSUNG
Intravesical oxybutynin is highly effective in the treatment of overactive bladder. Traditionally the mechanism of action was explained by antagonism of muscarinic receptors located in the detrusor, however evidence now suggests antimuscarinics may elicit their effect by modifying afferent pathways in the mucosal region. This study aimed to investigate the bladder wall distribution of oxybutynin in an ex vivo setting providing tissue - layer specific concentrations of drug achieved after intravesical delivery. Whole ex vivo porcine bladders were intravesically instilled with 0.167 mg mL(-1) oxybutynin solution. After 60 min, tissue samples were excised, serially sectioned parallel to the urothelial surface and extracted drug quantified. Drug distribution into the urothelium, lamina propria and detrusor was determined. Oxybutynin permeated into the bladder wall at a higher rate than other drugs previously investigated (apparent transurothelial Kp = 1.36 × 10(-5) cm s(-1) ). After 60 min intravesical instillation, concentrations achieved in the urothelium (298.69 μg g(-1) ) and lamina propria (43.65 μg g(-1) ) but not the detrusor (0.93 μg g(-1) ) were greater than reported IC50 values for oxybutynin. This work adds to the increasing body of evidence suggesting antimuscarinics elicit their effects via mechanisms other than direct inhibition of detrusor contraction.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Formaldehyd -Lösung, Molecular Biology, 36.5-38% in H2O
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Kaliumphosphat, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Formaldehyd -Lösung, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M
SAFC
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Kaliumphosphat, ReagentPlus®
Sigma-Aldrich
Formaldehyd -Lösung, Molecular Biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Kaliumphosphat, Molecular Biology, ≥98.0%
Sigma-Aldrich
Natriumchlorid, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Natriumchlorid -Lösung, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Natriumchlorid, 99.999% trace metals basis
Sigma-Aldrich
Formaldehyd -Lösung, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Acetonitril, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Stickstoff, ≥99.998%
Sigma-Aldrich
Kaliumphosphat, BioUltra, Molecular Biology, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Natriumchlorid, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Natriumchlorid -Lösung, 0.85%
Sigma-Aldrich
Kaliumphosphat, 99.99% trace metals basis
Sigma-Aldrich
Natriumchlorid-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Natriumchlorid, tablet
Sigma-Aldrich
Natriumchlorid, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Natriumchlorid, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Oxybutyninchlorid, ≥98% (TLC), powder