Merck
  • 18-β Glycyrrhetinic acid alleviates 2-acetylaminofluorene-induced hepatotoxicity in Wistar rats: Role in hyperproliferation, inflammation and oxidative stress.
18-β Glycyrrhetinic acid alleviates 2-acetylaminofluorene-induced hepatotoxicity in Wistar rats: Role in hyperproliferation, inflammation and oxidative stress.
Human & experimental toxicology (2014-10-30)
S K Hasan, R Khan, N Ali, A Q Khan, M U Rehman, M Tahir, A Lateef, S Nafees, S J Mehdi, S Rashid, A Shahid, S Sultana
ZUSAMMENFASSUNG
2-Acetylaminofluorene (2-AAF) is a known hepatic carcinogen which leads to tumour formation in rodents. 18-β Glycyrrhetinic acid (18β-GA) derived from liquorice plant has various pharmacological properties such as anti-ulcer, anti-inflammatory, antiviral, hepatoprotective and antioxidant. This study is designed to elucidate the chemopreventive properties of 18β-GA against 2-AAF-induced liver toxicity in Wistar rats and evaluated its effect on inflammatory and tumour promotion marker and activities of different oxidative stress enzymes. Administration of 2-AAF at the dose of (50 mg/kg body weight (b.w.) intraperitoneally (i.p.)) for five consecutive days induces hepatic toxicity, inflammation, oxidative stress and hyperproliferation. Pretreatment with 18β-GA at two different doses (45 and 75 mg kg(-1) b.w.) significantly ameliorates 2-AAF-induced increased lipid peroxidation, alanine transaminase and aspartate transaminase, xanthine oxidase activities and activities of phase-II detoxifying enzymes along with the levels of glutathione content. Administration of 18β-GA also significantly restored the expressions of proliferating cell nuclear antigen, cyclooxygenase 2, inducible nitric oxide synthase and nuclear factor κB. Furthermore, histological observations also support the preventive effects of 18β-GA. Our findings suggest that pretreatment with 18β-GA showed potential hepatoprotective effects via attenuation of oxidative stress, inflammation and hyperproliferation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
L-Glutathion reduziert, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
L-Glutathion reduziert, ≥98.0%
Sigma-Aldrich
Ethylendiamintetraessigsäure, 99.4-100.6%, powder
Sigma-Aldrich
Ethylendiamintetraessigsäure, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylendiamintetraessigsäure -Lösung, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Pyrogallol, ≥98% (HPLC)
Sigma-Aldrich
Pyrogallol, ACS reagent, ≥99%
Sigma-Aldrich
Ethylendiamintetraessigsäure, 99.995% trace metals basis
Sigma-Aldrich
Ethylendiamintetraessigsäure, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
2-Thiobarbitursäure, ≥98%
Sigma-Aldrich
1-Chlor-2,4-dinitrobenzol, ≥99%
Sigma-Aldrich
β-Nicotinamidadenin-Dinucleotidphosphat Hydrat
Sigma-Aldrich
1-Chlor-2,4-dinitrobenzol, 97%
Sigma-Aldrich
Xanthin, ≥99%
Sigma-Aldrich
L-Glutathion reduziert, BioXtra, ≥98.0%
Sigma-Aldrich
Ethylendiamintetraessigsäure, purified grade, ≥98.5%, powder
Sigma-Aldrich
Pyrogallol, ACS reagent
Sigma-Aldrich
Xanthin, ≥99.5% (HPLC), purified by recrystallization
Sigma-Aldrich
L-Glutathion, oxidiert Dinatriumsalz, ≥98%, powder
Sigma-Aldrich
2-Nitro-benzoesäure, 95%, Contains 3- and 4-isomers
Sigma-Aldrich
β-Nicotinamidadenin-Dinucleotidphosphat Natriumsalz, pkg of 5 mg (per vial)
Sigma-Aldrich
L-Glutathion, oxidiert Dinatriumsalz, suitable for cell culture, BioReagent
Sigma-Aldrich
Xanthin, BioUltra, ≥99%
Sigma-Aldrich
β-Nicotinamidadenin-Dinucleotidphosphat Natriumsalz, pkg of 10 mg (per vial)