Merck
  • RNA silencing of integrin-linked kinase increases the sensitivity of the A549 lung cancer cell line to cisplatin and promotes its apoptosis.
RNA silencing of integrin-linked kinase increases the sensitivity of the A549 lung cancer cell line to cisplatin and promotes its apoptosis.
Molecular medicine reports (2015-03-12)
Xiaozhen Zhao, Zhenye Xu, Zhongqi Wang, Zhonghua Wu, Yabin Gong, Lijuan Zhou, Yi Xiang
ZUSAMMENFASSUNG
The expression of integrin-linked kinase (ILK) has been reported to be involved in the regulation of integrin-mediated processes, including cancer cell proliferation, migration and invasion. Previous studies have demonstrated that inhibition of ILK may be an underlying approach for treating cancer. However, whether the knock down of ILK affects growth and apoptosis of lung cancer cells remains to be elucidated. Importantly, whether downregulation of ILK increases the sensitivity of lung cancer cells to cisplatin and amplifies cell apoptosis also remains to be elucidated. In the present study, ILK downregulation was mediated by lentivirus-mediated RNA interference. The expression levels of associated genes were determined by reverse-transcription quantitative polymerase chain reaction and western blotting. Cell proliferation was evaluated using a modified 3-(4,5-dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide assay and clone formation assay. The cell cycle and apoptosis were analyzed using flow cytometry. The current data revealed that lentivirus-mediated ILK gene silencing alone inhibited A549 cell proliferation and promotes cell cycle arrest, however, had no detectable effect on cell apoptosis. However, combined treatment with lentivirus-mediated ILK interference and cisplatin chemotherapy induced significantly more cell apoptosis than mono-chemotherapy or knockdown. The increased cell apoptosis and proliferation inhibition were attributed to abnormal downstream protein expression of ILK, including phospho-glycogen synthase kinase 3β, p-AKT, activator protein-1, β-catenin, cyclin D1 and matrix metalloproteinase-9. ILK inhibition may suppress the proliferation of A549 and increase A549 sensitivity to cisplatin. The combined treatment of ILK gene knockdown and chemotherapy has the potential to improve anticancer efficacy.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Natriumdodecylsulfat, BioReagent, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration)
Sigma-Aldrich
Natriumdodecylsulfat, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Natriumdodecylsulfat, ACS reagent, ≥99.0%
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Natriumdodecylsulfat, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Ethylendiamintetraessigsäure, 99.4-100.6%, powder
Sigma-Aldrich
Ethylendiamintetraessigsäure, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Ethylendiamintetraessigsäure -Lösung, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Natriumdodecylsulfat, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, 20% in H2O
SAFC
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
HEPES-Pufferlösung, 1 M in H2O
Supelco
Natriumdodecylsulfat, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Ethylendiamintetraessigsäure, 99.995% trace metals basis
Sigma-Aldrich
Ethylendiamintetraessigsäure, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Natriumorthovanadat, ≥90% (titration)
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
Natriumchlorid, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Natriumorthovanadat, 99.98% trace metals basis
Sigma-Aldrich
Natriumchlorid -Lösung, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Natriumchlorid, 99.999% trace metals basis