Merck
  • Rapid quick, easy, cheap, effective, rugged, and safe extraction with novel phospholipid cleanup: A streamlined ultra high performance liquid chromatography with ultraviolet detection approach for screening polycyclic aromatic hydrocarbons in avian blood cells and plasma.
Rapid quick, easy, cheap, effective, rugged, and safe extraction with novel phospholipid cleanup: A streamlined ultra high performance liquid chromatography with ultraviolet detection approach for screening polycyclic aromatic hydrocarbons in avian blood cells and plasma.
Journal of separation science (2015-05-28)
Anthony A Provatas, Alexander V Yevdokimov, Cory A King, Emma L Gatley, James D Stuart, David C Evers, Christopher R Perkins
ZUSAMMENFASSUNG
A streamlined method has been developed for the isolation and analysis of polycyclic aromatic hydrocarbons in avian blood cells and plasma utilizing quick, easy, cheap, effective, rugged, and safe extraction in combination with novel phospholipid cleanup technology. A variety of traditional extraction and cleanup techniques have been employed in the preparation and analysis of polycyclic aromatic hydrocarbonsin a variety of matrices; liquid-liquid partitioning, solid-phase extractions, gel permeation chromatography, and column chromatography are all effective techniques, however they are laborious and time consuming processes that require large amounts of solvent. Using quick, easy, cheap, effective, rugged, and safe extraction coupled with phospholipid cleanup, samples can be quickly screened while maintaining high throughput and sensitivity. With a liquid chromatography approach, analysis times may be kept short at 16 min while maintaining high analyte recovery. Recoveries in quality control samples ranged from 70 to 109%, with average surrogate recoveries of 80.6 ± 1.10%. The result of using a quick, easy, cheap, effective, rugged, and safe extraction approach in conjunction with phospholipid cleanup is a methodology that significantly reduces sample preparation time and solvent use while maintaining high sensitivity and reproducibility.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dichlormethan, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Natriumacetat, anhydrous, Molecular Biology, ≥99%
Sigma-Aldrich
Magnesiumsulfat, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Natriumacetat, 99.995% trace metals basis
Sigma-Aldrich
Natriumacetat, >99%, FG
Sigma-Aldrich
Magnesiumsulfat -Lösung, Molecular Biology, 1.00 M±0.04 M
Sigma-Aldrich
Acetonitril, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Natriumacetat, powder, BioReagent, suitable for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Naphthalin-d8, 99 atom % D, ≥98% (CP)
Sigma-Aldrich
Natriumacetat -Lösung, BioUltra, Molecular Biology, ~3 M in H2O
Sigma-Aldrich
Magnesiumsulfat -Lösung, BioUltra, Molecular Biology
Sigma-Aldrich
Natriumacetat, anhydrous, BioUltra, suitable for luminescence, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Ameisensäure, ≥95%, FCC, FG
Sigma-Aldrich
Magnesiumsulfat, ≥99.99% trace metals basis
Sigma-Aldrich
Natriumacetat, meets USP testing specifications, anhydrous
Sigma-Aldrich
Aceton, ≥99%, FCC, FG
Sigma-Aldrich
Dichlormethan, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Aceton, natural, ≥97%
Sigma-Aldrich
Aceton, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Chrysen-d12, 98 atom % D
Sigma-Aldrich
Natriumacetat, BioXtra, ≥99.0%
Sigma-Aldrich
Perylen-d12, 98 atom % D
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol -Lösung, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.