Merck
  • Adaptive optics stochastic optical reconstruction microscopy (AO-STORM) using a genetic algorithm.
Adaptive optics stochastic optical reconstruction microscopy (AO-STORM) using a genetic algorithm.
Optics express (2015-06-16)
Kayvan F Tehrani, Jianquan Xu, Yiwen Zhang, Ping Shen, Peter Kner
ZUSAMMENFASSUNG
The resolution of Single Molecule Localization Microscopy (SML) is dependent on the width of the Point Spread Function (PSF) and the number of photons collected. However, biological samples tend to degrade the shape of the PSF due to the heterogeneity of the index of refraction. In addition, there are aberrations caused by imperfections in the optical components and alignment, and the refractive index mismatch between the coverslip and the sample, all of which directly reduce the accuracy of SML. Adaptive Optics (AO) can play a critical role in compensating for aberrations in order to increase the resolution. However the stochastic nature of single molecule emission presents a challenge for wavefront optimization because the large fluctuations in photon emission do not permit many traditional optimization techniques to be used. Here we present an approach that optimizes the wavefront during SML acquisition by combining an intensity independent merit function with a Genetic algorithm (GA) to optimize the PSF despite the fluctuating intensity. We demonstrate the use of AO with GA in tissue culture cells and through ~50µm of tissue in the Drosophila Central Nervous System (CNS) to achieve a 4-fold increase in the localization precision.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natronlauge, BioUltra, Molecular Biology, 10 M in H2O
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natronlauge, 1.0 N, BioReagent, suitable for cell culture
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumhydroxid, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumhydroxid, ultra dry, powder or crystals, 99.99% trace metals basis
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
3-Ethyl-2,4-pentandion, Tautormermischung, 98%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumhydroxid-16O -Lösung, 20 wt. % in H216O, 99.9 atom % 16O
Preise und Verfügbarkeit sind derzeit nicht verfügbar.