Merck
  • High-throughput wide dynamic range procedure for the simultaneous quantification of nicotine and cotinine in multiple biological matrices using hydrophilic interaction liquid chromatography-tandem mass spectrometry.
High-throughput wide dynamic range procedure for the simultaneous quantification of nicotine and cotinine in multiple biological matrices using hydrophilic interaction liquid chromatography-tandem mass spectrometry.
Analytical and bioanalytical chemistry (2015-09-08)
Raúl Pérez-Ortuño, Jose M Martínez-Sánchez, Esteve Fernández, José A Pascual
ZUSAMMENFASSUNG
A straightforward, high-throughput method was developed and fully validated for the simultaneous determination of the specific tobacco biomarkers nicotine and its main metabolite cotinine in a wide dynamic range and supporting the most common human biological matrices (urine, oral fluid and hair). Sample preparation was performed inside the very HPLC injection vials by pipetting 0.5 mL of the liquid samples, deuterated internal standards in alkaline solution and dichloromethane as extraction solvent. Solid samples (i.e. around 10 mg hair) were first submitted to alkaline digestion in the HPLC vials and processed accordingly. The organic phase (reached through the upper aqueous layer) was directly injected without further treatment. Instrumental analysis was performed using hydrophilic interaction (HILIC) ultra-high performance liquid chromatography-tandem mass spectrometry (UHPLC-MS/MS). Total chromatographic time was 2 min. The method covers a wide dynamic range making it fit-for-purpose for the analysis of samples covering entire populations, irrespective of the level of exposure or tobacco use. Calibration curves (r (2) > 0.995) covered the range 1-2000 ng/mL (or 0.05-100 ng/mg hair) for nicotine and 0.1-2000 ng/mL (or 0.005-100 ng/mg hair) for cotinine. Within-run and between-run precision and accuracy were typically below 10 %, and always below 20 % at the lower limit of quantification. The method was successfully applied to the analysis of samples from different projects involving multiple matrices.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Dichlormethan, contains 40-150 ppm amylene as stabilizer, ACS reagent, ≥99.5%
Sigma-Aldrich
Kaliumchlorid, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Natriumhydroxid, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Ameisensäure, reagent grade, ≥95%
Sigma-Aldrich
Natriumhydroxid, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Natronlauge, 50% in H2O
Sigma-Aldrich
Acetonitril, ACS reagent, ≥99.5%
Sigma-Aldrich
Dichlormethan, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Ameisensäure, ACS reagent, ≥96%
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Natronlauge, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Ammoniak -Lösung, 7 N in methanol
Sigma-Aldrich
Natronlauge, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Kaliumchlorid, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Natriumhydroxid, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Kaliumchlorid, Molecular Biology, ≥99.0%
Sigma-Aldrich
Natriumhydroxid, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Kaliumchlorid, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Kaliumchlorid, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E508, 99-100.5% (AT), ≤0.0001% Al
Sigma-Aldrich
Ameisensäure, ACS reagent, ≥88%
Sigma-Aldrich
Natriumhydroxid, reagent grade, 97%, powder
Sigma-Aldrich
Dichlormethan, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Natriumhydroxid, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Kaliumchlorid, puriss. p.a., ≥99.5% (AT)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
(−)-Nikotin, ≥99% (GC), liquid