Merck
  • Repair synthesis step involving ERCC1-XPF participates in DNA repair of the Top1-DNA damage complex.
Repair synthesis step involving ERCC1-XPF participates in DNA repair of the Top1-DNA damage complex.
Carcinogenesis (2015-05-31)
Chiaki Takahata, Yuji Masuda, Arato Takedachi, Kiyoji Tanaka, Shigenori Iwai, Isao Kuraoka
ZUSAMMENFASSUNG
Topoisomerase 1 (Top1) is the intercellular target of camptothecins (CPTs). CPT blocks DNA religation in the Top1-DNA complex and induces Top1-attached nick DNA lesions. In this study, we demonstrate that excision repair cross complementing 1 protein-xeroderma pigmentosum group F (ERCC1-XPF) endonuclease and replication protein A (RPA) participate in the repair of Top1-attached nick DNA lesions together with other nucleotide excision repair (NER) factors. ERCC1-XPF shows nuclease activity in the presence of RPA on a 3'-phosphotyrosyl bond nick-containing DNA (Tyr-nick DNA) substrate, which mimics a Top1-attached nick DNA lesion. In addition, ERCC1-XPF and RPA form a DNA/protein complex on the nick DNA substrate in vitro, and co-localize in CPT-treated cells in vivo. Moreover, the DNA repair synthesis of Tyr-nick DNA lesions occurred in the presence of NER factors, including ERCC1-XPF, RPA, DNA polymerase delta, flap endonuclease 1 and DNA ligase 1. Therefore, some of the NER repair machinery might be an alternative repair pathway for Top1-attached nick DNA lesions. Clinically, these data provide insights into the potential of ERCC1 as a biomarker during CPT regimens.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Glycerin, Molecular Biology, ≥99.0%
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Phenylmethansulfonylfluorid, ≥98.5% (GC)
Sigma-Aldrich
Glycerin, ≥99.5%
Sigma-Aldrich
Ethylendiamintetraessigsäure, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Formamid, ≥99.5% (GC), BioReagent, Molecular Biology
Sigma-Aldrich
Glycerin -Lösung, 83.5-89.5% (T)
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Ethylendiamintetraessigsäure, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Formamid, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
Ethylendiamintetraessigsäure -Lösung, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Glycerin, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerin, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Glycerin, BioXtra, ≥99% (GC)
Sigma-Aldrich
Phenylmethansulfonylfluorid, ≥99.0% (T)
Sigma-Aldrich
Ethylendiamintetraessigsäure, 99.995% trace metals basis
Sigma-Aldrich
Ethylendiamintetraessigsäure, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Bromphenolblau, Sulton Form, ACS reagent
Sigma-Aldrich
Glycerin, FCC, FG
Sigma-Aldrich
Bromphenolblau, titration: suitable
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Dimethylsulfoxid, PCR Reagent
Sigma-Aldrich
o-Xylen, anhydrous, 97%
Sigma-Aldrich
Ethylendiamintetraessigsäure, purified grade, ≥98.5%, powder
Sigma-Aldrich
Formamid -Lösung, suitable for NMR (reference standard), 90% in DMSO-d6 (99.9 atom % D), NMR tube size 10 mm × 8 in.