Merck
  • Influence of a survivin suppressor YM155 on the chemoresistance of canine histiocytic sarcoma cells.
Influence of a survivin suppressor YM155 on the chemoresistance of canine histiocytic sarcoma cells.
Veterinary journal (London, England : 1997) (2015-06-07)
Hiroki Yamazaki, Satoshi Takagi, Kenji Hosoya, Masahiro Okumura
ZUSAMMENFASSUNG
Histiocytic sarcoma (HS) in dogs exhibits aggressive biological behaviors and currently few effective treatments are available. Survivin could serve as a potential therapeutic target in several cancers. Sepantronium bromide (YM155) is a potential novel survivin-targeting agent and in this study the influence of survivin expression on clinical outcomes and the effects of YM155 on biological activities in HS cells were investigated. Specimens of HS dogs (n = 30) and four canine HS cell lines were used. The correlation between survivin expression and clinical outcome in the HS dogs was retrospectively assessed using quantitative PCR. Following YM155 treatment of cell lines, apoptosis, cell viability, and drug transporter activities were evaluated using annexin V staining, methylthiazole tetrazolium assays, and Hoechst-33342 staining, respectively. Elevated survivin expression in the HS dogs corresponded with reduced disease-free intervals and survival time, and increased chemoresistance, which led to poor clinical outcomes. Furthermore, YM155 treatment suppressed cell-growth and resistance to lomustine in HS cells by inhibiting the activity of ATP-binding cassette transporters. The evidence presented here supports favorable preclinical evaluation and indicates that survivin-targeted therapies might be effective against HS dogs.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Natriumazid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Natriumazid, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Natriumazid, purum p.a., ≥99.0% (T)
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Natriumazid, BioXtra
Sigma-Aldrich
Dimethylsulfoxid, PCR Reagent
Sigma-Aldrich
Desoxyribonuklease I Rind, recombinant, expressed in Pichia pastoris, buffered aqueous glycerol solution, ≥5,000 units/mg protein
Sigma-Aldrich
Desoxyribonuklease I Rind, recombinant, expressed in Pichia pastoris, lyophilized powder, RNAse and protease, free
Sigma-Aldrich
Tris(tert-butoxy)silanol, 99.999%
Sigma-Aldrich
Dimethylsulfoxid, ≥99.6%, ReagentPlus®