Merck
  • Preparation and Characterization of Dictyophora indusiata Polysaccharide-Zinc Complex and Its Augmented Antiproliferative Activity on Human Cancer Cells.
Preparation and Characterization of Dictyophora indusiata Polysaccharide-Zinc Complex and Its Augmented Antiproliferative Activity on Human Cancer Cells.
Journal of agricultural and food chemistry (2015-07-15)
Wenzhen Liao, Yunjun Lu, Junning Fu, Zhengxiang Ning, Jiguo Yang, Jiaoyan Ren
ZUSAMMENFASSUNG
Dictyophora indusiata polysaccharide (DP1) was successfully chelated with zinc chloride to achieve its enhanced antiproliferative activity. The obtained DP1-Zn complex showed significant antiproliferative activity (18.1 ± 2.84% viability of MCF-7 cells at 250 μg/mL) on a group of human cancer cell lines through induction of apoptosis. The pro-apoptotic action of DP1-Zn was confirmed by morphological changes including chromatin condensation, DNA breakage, and S phase cell cycle arrest in human breast adenocarcinoma cells (MCF-7). The DP1-Zn-induced apoptotic pathways were characterized by the activation of caspases-3, -8, and -9, mitochondrial dysfunction, and reactive oxygen species (ROS) overproduction (305 ± 7.06% production of control at 250 μg/mL). This study suggested that DP1-Zn can be developed as a candidate for cancer treatment and prevention, especially human breast adenocarcinoma.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Diethylpyrocarbonat, 96% (NT)
Sigma-Aldrich
Thiazolylblau-Tetrazoliumbromid, 98%
Sigma-Aldrich
L-Ascorbinsäure, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
Thiazolylblau-Tetrazoliumbromid, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
L-Ascorbinsäure, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbinsäure, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbinsäure, 99%
Sigma-Aldrich
L-Ascorbinsäure, reagent grade, crystalline
Sigma-Aldrich
Natriumselenit, 99%
Sigma-Aldrich
Natriumselenit, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
Natriumselenit, γ-irradiated, lyophilized powder, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Ascorbinsäure, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbinsäure, meets USP testing specifications
Sigma-Aldrich
L-Ascorbinsäure, reagent grade
Sigma-Aldrich
Diethylpyrocarbonat, 96% (NT)
Sigma-Aldrich
Diethylpyrocarbonat, 96% (NT)
Sigma-Aldrich
L-Ascorbinsäure, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbinsäure, FCC, FG
Sigma-Aldrich
L-Ascorbinsäure, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
JC-1, powder or solid (Crystals)
Sigma-Aldrich
L-Ascorbinsäure, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
2,2′-Bichinolin-4,4′-dicarbonsäure Dinatriumsalz Dihydrat, ≥98% (HPLC)
Sigma-Aldrich
N-Acetyl-Leu-Glu-His-Asp-7-amido-4-trifluoromethylcoumarin, ≥95% (HPLC), powder