Merck
Chloroquine and Hydroxychloroquine Increase Retinal Pigment Epithelial Layer Permeability.
Journal of biochemical and molecular toxicology (2015-03-11)
Nicoline M Korthagen, Jeroen Bastiaans, Jan C van Meurs, Kiki van Bilsen, P Martin van Hagen, Willem A Dik
ZUSAMMENFASSUNG
Antimalarials chloroquine (CQ) and hydroxychloroquine (HCQ) are widely used as antiinflammatory drugs, but side effects include retinopathy and vision loss. The objective of this study was to examine the effect of CQ and HCQ on the barrier integrity of retinal pigment epithelial (RPE) cell monolayers in vitro. Permeability of ARPE-19 cell monolayers was determined using Fluorescein isothiocyanate (FITC)-labeled dextran. The influence of CQ and HCQ on cell death and the expression tight junction molecules was examined. CQ and HCQ significantly increased ARPE-19 monolayer permeability after 3 and 18 h, respectively, and enhanced mRNA levels for claudin-1 and occludin. Cytotoxicity was only observed after 18 h exposure. Thus, CQ and HCQ rapidly enhance RPE barrier permeability in vitro, independent of cytotoxicity or loss of zonula occludens-1, claudin-1, and occludin expression. Our findings suggest that CQ/HCQ-induced permeability of the RPE layer may contribute to blood-retinal barrier breakdown in case of CQ/HCQ-induced retinopathy.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Glycerin, Molecular Biology, ≥99.0%
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
L-Glutamin, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
Glycerin, ≥99.5%
Sigma-Aldrich
L-Glutamin, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Glycerin -Lösung, 83.5-89.5% (T)
Sigma-Aldrich
Glycerin, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Fluoreszein-5(6)-isothiocyanat, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
Glycerin, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
SAFC
L-Glutamin
Sigma-Aldrich
Glycerin, BioXtra, ≥99% (GC)
Sigma-Aldrich
HEPES-Pufferlösung, 1 M in H2O
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
Glycerin, FCC, FG
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
L-Glutamin, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
Ethansulfonsäure, 95%
Sigma-Aldrich
Fluoreszein-5(6)-isothiocyanat, ≥90% (HPLC)
Sigma-Aldrich
Fluorescein-Isothiocyanat Isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
L-Glutamin, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamin
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%