Merck
  • The anthelmintic efficacy of natural plant cysteine proteinases against Hymenolepis microstoma in vivo.
The anthelmintic efficacy of natural plant cysteine proteinases against Hymenolepis microstoma in vivo.
Journal of helminthology (2014-09-17)
F Mansur, W Luoga, D J Buttle, I R Duce, A Lowe, J M Behnke
ZUSAMMENFASSUNG
Little is known about the efficacy of cysteine proteinases (CP) as anthelmintics for cestode infections in vivo. Hymenolepis microstoma is a natural parasite of house mice, and provides a convenient model system for the assessment of novel drugs for anthelmintic activity against cestodes. The experiments described in this paper indicate that treatment of H. microstoma infections in mice with the supernatant of papaya latex (PLS), containing active cysteine proteinases, is only minimally efficacious. The statistically significant effects seen on worm burden and biomass showed little evidence of dose dependency, were temporary and the role of cysteine proteinases as the active principles in PLS was not confirmed by specific inhibition with E-64. Worm fecundity was not affected by treatment at the doses used. We conclude also that this in vivo host-parasite system is not sensitive enough to be used reliably for the detection of cestocidal activity of compounds being screened as potential, novel anthelmintics.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
L-Cystein, from non-animal source, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
L-Cystein, 97%
Sigma-Aldrich
Glutaraldehyd -Lösung, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Sigma-Aldrich
Glutaraldehyd -Lösung, Grade II, 25% in H2O
Sigma-Aldrich
Glutaraldehyd -Lösung, 50 wt. % in H2O
Sigma-Aldrich
E-64, protease inhibitor
Sigma-Aldrich
Nα-Benzoyl-DL-Arginin-4-Nitroanilid -hydrochlorid, ≥98%
Sigma-Aldrich
L-Cystein, BioUltra, ≥98.5% (RT)
Sigma-Aldrich
Fluorethylencarbonat, 99%
Sigma-Aldrich
Glutaraldehyd -Lösung, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
SAFC
L-Cystein
Sigma-Aldrich
L-Cystein, ≥97%, FG
Sigma-Aldrich
Glutaraldehyd -Lösung, Grade I, 70% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutarsäure-Dialdehyd -Lösung, 50 wt. % in H2O, FCC
Sigma-Aldrich
Glutaraldehyd -Lösung, Grade I, 8% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyd -Lösung, 50% in H2O, suitable for photographic applications
Sigma-Aldrich
L-Cystein, Wacker Chemie AG, ≥98.0%