Merck
  • Activation of autophagy in human skeletal muscle is dependent on exercise intensity and AMPK activation.
Activation of autophagy in human skeletal muscle is dependent on exercise intensity and AMPK activation.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2015-05-10)
Céline Schwalm, Cécile Jamart, Nicolas Benoit, Damien Naslain, Christophe Prémont, Jérémy Prévet, Ruud Van Thienen, Louise Deldicque, Marc Francaux
ZUSAMMENFASSUNG
In humans, nutrient deprivation and extreme endurance exercise both activate autophagy. We hypothesized that cumulating fasting and cycling exercise would potentiate activation of autophagy in skeletal muscle. Well-trained athletes were divided into control (n = 8), low-intensity (LI, n = 8), and high-intensity (HI, n = 7) exercise groups and submitted to fed and fasting sessions. Muscle biopsy samples were obtained from the vastus lateralis before, at the end, and 1 h after a 2 h LI or HI bout of exercise. Phosphorylation of ULK1(Ser317) was higher after exercise (P < 0.001). In both the fed and the fasted states, LC3bII protein level and LC3bII/I were decreased after LI and HI (P < 0.05), while p62/SQSTM1 was decreased only 1 h after HI (P < 0.05), indicating an increased autophagic flux after HI. The autophagic transcriptional program was also activated, as evidenced by the increased level of LC3b, p62/SQSTM1, GabarapL1, and Cathepsin L mRNAs observed after HI but not after LI. The increased autophagic flux after HI exercise could be due to increased AMP-activated protein kinase α (AMPKα) activity, as both AMPKα(Thr172) and ACC(Ser79) had a higher phosphorylation state after HI (P < 0.001). In summary, the most effective strategy to activate autophagy in human skeletal muscle seems to rely on exercise intensity more than diet.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Saccharose, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Saccharose, ≥99.5% (GC)
Sigma-Aldrich
DL-Dithiothreitol -Lösung, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Saccharose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Saccharose, BioUltra, Molecular Biology, ≥99.5% (HPLC)
Supelco
DL-Dithiothreitol -Lösung, 1 M in H2O
Sigma-Aldrich
Ethylendiamintetraessigsäure, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenglykol-bis(2-aminoethylether)-N,N,N′,N′-Tetraessigsäure, Molecular Biology, ≥97.0%
Sigma-Aldrich
Ethylendiamintetraessigsäure, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylendiamintetraessigsäure -Lösung, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Saccharose, ≥99.5% (GC)
Sigma-Aldrich
Saccharose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Saccharose, ACS reagent
Sigma-Aldrich
Natriumpyrophosphat, ≥95%
Sigma-Aldrich
Ethylendiamintetraessigsäure, 99.995% trace metals basis
Sigma-Aldrich
Ethylendiamintetraessigsäure, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Natriumorthovanadat, ≥90% (titration)
Sigma-Aldrich
Lidocain, powder
Sigma-Aldrich
Anti-LC3B in Kaninchen hergestellte Antikörper, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Ethylenglykol-bis(2-aminoethylether)-N,N,N′,N′-Tetraessigsäure, ≥97.0%
Sigma-Aldrich
Natriumorthovanadat, 99.98% trace metals basis
Sigma-Aldrich
Saccharose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sigma-Aldrich
Saccharose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Natriumfluorid, 99.99% trace metals basis
Sigma-Aldrich
DL-Glyceraldehyd-3-phosphat -Lösung, 45-55 mg/mL in H2O
Sigma-Aldrich
Ethylendiamintetraessigsäure, purified grade, ≥98.5%, powder
Sigma-Aldrich
Anti-p62/SQSTM1 in Kaninchen hergestellte Antikörper, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Saccharose, ≥99% (GC), Grade I, suitable for plant cell culture
Sigma-Aldrich
Lidocain, analytical standard
Sigma-Aldrich
Saccharose, meets USP testing specifications