Merck
Phytase activity in lichens.
The New phytologist (2015-05-13)
Niall F Higgins, Peter D Crittenden
ZUSAMMENFASSUNG
Phytase activity was investigated in 13 lichen species using a novel assay method. The work tested the hypothesis that phytase is a component of the suite of surface-bound lichen enzymes that hydrolyse simple organic forms of phosphorus (P) and nitrogen (N) deposited onto the thallus surface. Hydrolysis of inositol hexaphosphate (InsP6 , the substrate for phytase) and appearance of lower-order inositol phosphates (InsP5 -InsP1 ), the hydrolysis products, were measured by ion chromatography. Phytase activity in Evernia prunastri was compared among locations with contrasting rates of N deposition. Phytase activity was readily measurable in epiphytic lichens (e.g. 11.3 μmol InsP6 hydrolysed g(-1)  h(-1) in Bryoria fuscescens) but low in two terricolous species tested (Cladonia portentosa and Peltigera membranacea). Phytase and phosphomonoesterase activities were positively correlated amongst species. In E. prunastri both enzyme activities were promoted by N enrichment and phytase activity was readily released into thallus washings. InsP6 was not detected in tree canopy throughfall but was present in pollen leachate. Capacity to hydrolyse InsP6 appears widespread amongst lichens potentially promoting P capture from atmospheric deposits and plant leachates, and P cycling in forest canopies. The enzyme assay used here might find wider application in studies on plant root-fungal-soil systems.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumhydroxid, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Kaliumphosphat, ACS reagent, ≥98%
Sigma-Aldrich
Natriumhydroxid, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Natronlauge, 50% in H2O
Sigma-Aldrich
Kaliumnitrat, ACS reagent, ≥99.0%
Sigma-Aldrich
Natronlauge, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Natronlauge, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Natriumhydroxid, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Natriumhydroxid, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Ethylendiamintetraessigsäure, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Natriumhydroxid, reagent grade, 97%, powder
Sigma-Aldrich
Natriumhydroxid, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Ethylendiamintetraessigsäure, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Ethylendiamintetraessigsäure -Lösung, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Kaliumnitrat, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99%
SAFC
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Kaliumnitrat, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Natronlauge, 5.0 M
Sigma-Aldrich
Natriumhydroxid, beads, 16-60 mesh, reagent grade, 97%
Sigma-Aldrich
Ethylendiamintetraessigsäure, 99.995% trace metals basis
Sigma-Aldrich
Ethylendiamintetraessigsäure, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Kaliumphosphat, anhydrous, suitable for luminescence, Molecular Biology, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Natriumchlorid, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Natriumhydroxid, reagent grade, 97%, flakes