Merck
  • Ovatodiolide sensitizes aggressive breast cancer cells to doxorubicin, eliminates their cancer stem cell-like phenotype, and reduces doxorubicin-associated toxicity.
Ovatodiolide sensitizes aggressive breast cancer cells to doxorubicin, eliminates their cancer stem cell-like phenotype, and reduces doxorubicin-associated toxicity.
Cancer letters (2015-05-16)
Oluwaseun Adebayo Bamodu, Wen-Chien Huang, David T W Tzeng, Alexander Wu, Liang Shun Wang, Chi-Tai Yeh, Tsu-Yi Chao
ZUSAMMENFASSUNG
Triple-negative breast cancer (TNBC) is chemotherapy-refractory and associated with poor clinical prognosis. Doxorubicin (Doxo), a class I anthracycline and first-line anticancer agent, effective against a wide spectrum of neoplasms including breast carcinoma, is associated with several cumulative dose-dependent adverse effects, including cardiomyopathy, typhilitis, and acute myelotoxicity. This study evaluated the usability of Ovatodiolide (Ova) in sensitizing TNBC cells to Doxo cytotoxicity, so as to reduce Doxo effective dose and consequently its adverse effects. TNBC cell lines MDA-MB-231 and HS578T were used. Pre-treatment of the TNBC cells with 10 µM Ova 24 h before Doxo administration increased the Doxo anticancer effect (IC50 1.4 µM) compared to simultaneous treatment with Doxo ( IC50 1.8 µM), or Doxo alone (IC50 9.2 µM). Intracellular accumulation of Doxo was lowest in Ova pre-treated cells at all Doxo concentrations, when compared with Doxo or simultaneously treated cells. In comparison to the Doxo-only group, cell cycle analysis of MDA-MB-231 cells treated concurrently with 2.5 µM Ova and 1.25 µM Doxo showed increased percentage of cells arrested at G0/G1; however, pre-treatment with the same concentration of Ova 24 h before Doxo showed greater tumor growth inhibition, with a 2.4-fold increased percentage of cells in G0/G1 arrest, greater Doxo-induced apoptosis, and significantly reduced intracellular Doxo accumulation. Additionally, Ova-sensitized TNBC cells also lost their cancer stem cell-like phenotype evidenced by significant dissolution, necrosis of formed mammospheres. Taken together, these findings indicate that Ova sensitizes TNBC cells to Doxo and potentiates doxorubicin-induced elimination of the TNBC cancer stem cell-like phenotype.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Doxorubicin -hydrochlorid, 98.0-102.0% (HPLC)
Sigma-Aldrich
Ethylendiamintetraessigsäure, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Ethylendiamintetraessigsäure, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylendiamintetraessigsäure -Lösung, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylendiamintetraessigsäure, 99.995% trace metals basis
Sigma-Aldrich
Ethylendiamintetraessigsäure, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Doxorubicin -hydrochlorid, suitable for fluorescence, 98.0-102.0% (HPLC)
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Essigsäure, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Dimethylsulfoxid, PCR Reagent
Sigma-Aldrich
5α-Androstan-17β-ol-3-on, ≥97.5%
Sigma-Aldrich
Sulforhodamin B Mononatriumsalz, Dye content 75 %
Sigma-Aldrich
Sulforhodamin B Natriumsalz, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Essigsäure, ≥99.5%, FCC, FG
Sigma-Aldrich
Ethylendiamintetraessigsäure, purified grade, ≥98.5%, powder
Sigma-Aldrich
Essigsäure, natural, ≥99.5%, FG
Sigma-Aldrich
Sulforhodamin B Natriumsalz, Technical grade
Sigma-Aldrich
5α-Androstan-17β-ol-3-on, purum, ≥99.0% (TLC)
Sigma-Aldrich
8-Octanoyloxypyren-1,3,6-Trisulfonsäure Trinatriumsalz, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Essigsäure-12C2, 99.9 atom % 12C
Sigma-Aldrich
Dimethylsulfoxid, ≥99.6%, ReagentPlus®