Merck
  • The effect of thiacloprid formulation on DNA/chromosome damage and changes in GST activity in bovine peripheral lymphocytes.
The effect of thiacloprid formulation on DNA/chromosome damage and changes in GST activity in bovine peripheral lymphocytes.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes (2015-08-15)
Martina Galdíková, Katarína Šiviková, Beáta Holečková, Ján Dianovský, Monika Drážovská, Viera Schwarzbacherová
ZUSAMMENFASSUNG
The potential genotoxic effect of thiacloprid formulation on bovine peripheral lymphocytes was evaluated using the comet assay and the cytogenetic endpoints: chromosome aberrations (CAs), sister chromatid exchanges (SCEs) and micronuclei (MNi). Whole blood cultures were treated with the insecticide at concentrations of 30, 60, 120, 240 and 480 μg mL(-1) for 24, 48 h and/or 2 h of incubation. A statistically significant increase in the frequency of DNA damage, as well as in unstable chromosome aberrations (% breaks) were found after exposure to the insecticide at concentrations ranging from 120 to 480 μg mL(-1) (P < 0.05, P < 0.01, P < 0.001). For the detection of stable structural chromosome aberrations (e.g., translocations) and numerical aberrations by the FISH method, three whole chromosome painting probes for bovine chromosomes 1, 5 and 7 (BTA1, BTA5 and BTA7) were used in our experiments. We observed numerical aberrations, but without any statistical significance. Regarding the sister chromatid exchanges, no significant elevation in the SCE frequencies was found after 24-h exposure to the insecticide. A dose-related response in the SCE induction was obtained in bovine cultures after the prolonged time of exposure (48 h) to thiacloprid formulation at concentrations ranging from 120 to 480 μg mL(-1) in each donor (P < 0.05, P < 0.01), which was associated with a reduction of the PI (P < 0.05, P < 0.01). The insecticide failed to produce MNi; however, a significant reduction of CBPI was observed. Using real-time PCR, a decrease in the expression of bovine glutathione S-transferase M3 (GSTM3) was detected at the lowest dose. The higher concentrations of thiacloprid formulation caused an increase in the mRNA expression.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Wasserstoffperoxid -Lösung, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration)
Sigma-Aldrich
L-Glutamin, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
Mitomycin C aus Streptomyces caespitosus, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
L-Glutamin, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Colchicin, ≥95% (HPLC), powder
Sigma-Aldrich
Ethylmethansulfonat, liquid
SAFC
L-Glutamin
Sigma-Aldrich
HEPES-Pufferlösung, 1 M in H2O
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
Colchicin, suitable for plant cell culture, BioReagent, ≥95% (HPLC)
Sigma-Aldrich
Mitomycin C aus Streptomyces caespitosus, powder, contains NaCl as solubilizer
Sigma-Aldrich
Wasserstoffperoxid -Lösung, 34.5-36.5%
Sigma-Aldrich
L-Glutamin, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
Mitomycin C aus Streptomyces caespitosus, ≥98% (HPLC), γ-irradiated, suitable for cell culture
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
Mitomycin C aus Streptomyces caespitosus, meets USP testing specifications
Sigma-Aldrich
L-Glutamin, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamin
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
Magnesium-methylcarbonat -Lösung, 2.0 M in DMF