Merck
  • Determination of polycyclic aromatic hydrocarbons using lab on valve dispersive liquid-liquid microextraction coupled to high performance chromatography.
Determination of polycyclic aromatic hydrocarbons using lab on valve dispersive liquid-liquid microextraction coupled to high performance chromatography.
Talanta (2015-04-13)
M Fernández, S Clavijo, R Forteza, V Cerdà
ZUSAMMENFASSUNG
In this work, dispersive liquid-liquid microextraction (DLLME) method was applied for high performance liquid chromatography (HPLC) determination of 15 PAHs in aqueous matrices.The extraction procedure was automated using a system of multisyringe flow injection analysis coupled to HPLC instrument with fluorescence detector. Factors affecting the extraction process, such as type and volume of extraction and dispersive solvent, extraction time and centrifugation step were investigated thoroughly and optimized utilizing factorial design. The best recovery was achieved using 100 µL of trichloroethylene as the extraction solvent and 900 µL of acetonitrile as the dispersive solvent.The results showed that extraction time has no effect on the recovery of PAHs. The enrichment factors of PAHs were in the range of 86-95 with limits of detection of 0.02-0.6 µg L(-1). The linearity was 0.2-600 µg L(-1) for different PAHs. The relative standard deviation (RSD) for intra- and inter-day of extraction of PAHs were in the range of 1.6-4.7 and 2.1-5.3, respectively, for five measurements.The developed method was used to assess the occurrence of 15 PAHs in tap water, rain waters and river surface waters samples.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dichlormethan, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Benzo[a]pyren, ≥96% (HPLC)
Sigma-Aldrich
Naphthalin, 99%
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Pyren, 98%
Sigma-Aldrich
Anthracen, ReagentPlus®, 99%
Sigma-Aldrich
Fluoren, 98%
Sigma-Aldrich
Phenanthren, 98%
Sigma-Aldrich
Acetonitril, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Anthracen, reagent grade, 97%
Sigma-Aldrich
Acenaphthen, 99%
Sigma-Aldrich
Naphthalin, suitable for scintillation, ≥99%
Sigma-Aldrich
Pyren, puriss. p.a., suitable for fluorescence, ≥99.0% (GC)
Sigma-Aldrich
Aceton, ≥99%, FCC, FG
Sigma-Aldrich
Benz[a]anthracen, 99%
Sigma-Aldrich
Dichlormethan, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Fluoranthen, 98%
Sigma-Aldrich
Aceton, natural, ≥97%
Sigma-Aldrich
Benzo[b]fluoranthen, 98%
Sigma-Aldrich
Acenaphthylin, 99%
Sigma-Aldrich
Aceton, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Trichlorethylen, anhydrous, contains 40 ppm diisopropylamine as stabilizer, ≥99%
Sigma-Aldrich
Phenanthren, sublimed grade, ≥99.5%
Sigma-Aldrich
Benzo[k]fluoranthen, suitable for fluorescence, ≥99%
Sigma-Aldrich
Anthracen, suitable for scintillation, ≥99.0% (GC)
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)