Merck
  • Development and characterization of a ricinoleic acid poloxamer gel system for transdermal eyelid delivery.
Development and characterization of a ricinoleic acid poloxamer gel system for transdermal eyelid delivery.
Drug development and industrial pharmacy (2014-02-13)
Sai H S Boddu, Sindhu Prabha Bonam, Rose Jung
ZUSAMMENFASSUNG
This study deals with the preparation and evaluation of a pluronic lecithin organogel (PLO gel) containing ricinoleic acid for the transdermal eyelid delivery of dexamethasone and tobramycin. Five different PLO gel formulations (F1, F2, F3, F4 and F5) containing tobramycin (0.3%) and dexamethasone (0.1%) were prepared and compared to a conventional PLO gel (light mineral oil PLO gel, F6) with respect to physical appearance and viscosity. The optimized ricinoleic acid PLO gel formulation (F2) was further characterized for pH, gelation temperature, morphology and drug content. Ex vivo permeability of dexamethasone and bactericidal activity of tobramycin from formulation F2 was tested, and values were compared to the marketed Tobradex® eye ointment. No apparent changes in the physical appearance and consistency were observed when ricinoleic acid was used as the oil phase. The pH of the optimized ricinoleic acid PLO gel (formulation F2) was found to be 6.54 with a gelation temperature of 31 °C. The drug content of tobramycin and dexamethasone were found to be 102.8% and 100.14%, respectively. The penetration profile of dexamethasone from formulation F2 was found to be much higher than the marketed Tobradex® eye ointment. F2 showed a better antimicrobial activity and higher zones of inhibition when compared to the marketed Tobradex® eye ointment. The findings of this investigation indicate that the ricinoleic acid PLO gel has the potential for use as a transdermal eyelid delivery system.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M
SAFC
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Natriumchlorid, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Natriumchlorid -Lösung, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Natriumchlorid, 99.999% trace metals basis
Sigma-Aldrich
Acetonitril, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Rizinolsäure, ≥95%
Sigma-Aldrich
Tobramycin, Aminoglycoside antibiotic
Sigma-Aldrich
Natriumchlorid, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Natriumchlorid -Lösung, 0.85%
Sigma-Aldrich
Natriumchlorid, tablet
Sigma-Aldrich
Natriumchlorid-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Natriumchlorid, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Natriumchlorid, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol -Lösung, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.