Merck
Thioredoxin-interacting protein regulates the differentiation of murine erythroid precursors.
Experimental hematology (2015-01-21)
Jadwiga J Gasiorek, Marc Mikhael, Daniel Garcia-Santos, Simon T Hui, Prem Ponka, Volker Blank
ZUSAMMENFASSUNG
Thioredoxin-interacting protein (TXNIP) is involved in various cellular processes including redox control, metabolism, differentiation, growth, and apoptosis. With respect to hematopoiesis, TXNIP has been shown to play roles in natural killer cells, dendritic cells, and hematopoietic stem cells. Our study investigates the role of TXNIP in erythropoiesis. We observed a rapid and significant increase of TXNIP transcript and protein levels in mouse erythroleukemia cells treated with dimethyl sulfoxide or hexamethylene bisacetamide, inducers of erythroid differentiation. The upregulation of TXNIP was not abrogated by addition of the antioxidant N-acetylcysteine. The increase of TXNIP expression was confirmed in another model of erythroid differentiation, G1E-ER cells, which undergo differentiation upon activation of the GATA1 transcription factor. In addition, we showed that TXNIP levels are induced following inhibition of p38 or c-Jun N-terminal kinase (JNK) mitogen-activated protein kinases. We also observed an increase in iron uptake and a decrease in transferrin receptor protein upon TXNIP overexpression, suggesting a role in iron homeostasis. In vivo, flow cytometry analysis of cells from Txnip(-/-) mice revealed a new phenotype of impaired terminal erythropoiesis in the spleen, characterized by a partial block between basophilic and late basophilic/polychromatic erythroblasts. Based on our data, TXNIP emerges as a novel regulator of terminal erythroid differentiation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Natriumdodecylsulfat, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
N-Acetyl-L-Cystein, BioReagent, suitable for cell culture
Sigma-Aldrich
Natriumdodecylsulfat, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Natriumdodecylsulfat, ACS reagent, ≥99.0%
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
N-Acetyl-L-Cystein, Sigma Grade, ≥99% (TLC), powder
Sigma-Aldrich
Natriumdodecylsulfat, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
1-Thioglycerin, liquid, BioReagent, suitable for cell culture, ≥97% (titration)
Sigma-Aldrich
Natriumdodecylsulfat, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, 20% in H2O
Sigma-Aldrich
Fluoreszein-5(6)-isothiocyanat, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Supelco
Natriumdodecylsulfat, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
1-Thioglycerin, ≥99.0% (GC)
Sigma-Aldrich
1-Thioglycerin, ≥97%
Sigma-Aldrich
Natriumdodecylsulfat, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Dimethylsulfoxid, PCR Reagent
Sigma-Aldrich
Natriumdodecylsulfat, ≥98.0% (GC)
Sigma-Aldrich
Natriumdodecylsulfat, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
N-Acetyl-L-Cystein, BioXtra, ≥99% (TLC)
Sigma-Aldrich
Fluoreszein-5(6)-isothiocyanat, ≥90% (HPLC)
Sigma-Aldrich
SP600125, ≥98% (HPLC)
Sigma-Aldrich
Fluorescein-Isothiocyanat Isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Natriumdodecylsulfat, ≥90% ((Assay))