Merck
Inhibition of insulin fibrillation by osmolytes: Mechanistic insights.
Scientific reports (2015-12-01)
Sinjan Choudhary, Nand Kishore, Ramakrishna V Hosur
ZUSAMMENFASSUNG
We have studied here using a number of biophysical tools the effects of osmolytes, betaine, citrulline, proline and sorbitol which differ significantly in terms of their physical characteristics such as, charge distribution, polarity, H-bonding abilities etc, on the fibrillation of insulin. Among these, betaine, citrulline, and proline are very effective in decreasing the extent of fibrillation. Proline also causes a substantial delay in the onset of fibrillation in the concentration range (50-250 mM) whereas such an effect is seen for citrulline only at 250 mM, and in case of betaine this effect is not seen at all in the whole concentration range. The enthalpies of interaction at various stages of fibrillation process have suggested that the preferential exclusion of the osmolyte and its polar interaction with the protein are important in inhibition. The results indicate that the osmolytes are most effective when added prior to the elongation stage of fibrillation. These observations have significant biological implications, since insulin fibrillation is known to cause injection amyloidosis and our data may help in designing lead drug molecules and development of potential therapeutic strategies.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
D-Sorbit, ≥98% (GC)
Sigma-Aldrich
Betain -Lösung, 5 M, PCR Reagent
Sigma-Aldrich
L-Prolin, from non-animal source, meets EP, USP testing specifications, suitable for cell culture
Sigma-Aldrich
Betain, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
D-Sorbit, BioUltra, ≥99.0% (HPLC)
Sigma-Aldrich
Betain, ≥98% (perchloric acid titration)
Sigma-Aldrich
Thioflavin T, used as stain for amyloid
Sigma-Aldrich
D-Sorbit, 99% (GC)
Sigma-Aldrich
L-Prolin, ReagentPlus®, ≥99% (HPLC)
SAFC
L-Prolin
Sigma-Aldrich
D-Sorbit, ≥98% (GC), Molecular Biology
Sigma-Aldrich
L-Prolin, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
D-Sorbit, ≥98% (GC), BioReagent, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
L-Prolin, 99%, FCC, FG
Sigma-Aldrich
Sorbit F -Lösung, 70 wt. % in H2O, Contains mainly D-sorbitol with lesser amounts of other hydrogenated oligosaccharides
Sigma-Aldrich
D-Sorbit, ≥98% (GC), BioXtra
Sigma-Aldrich
D-Sorbit, FCC, FG