Merck
Defining human ERAD networks through an integrative mapping strategy.
Nature cell biology (2011-11-29)
John C Christianson, James A Olzmann, Thomas A Shaler, Mathew E Sowa, Eric J Bennett, Caleb M Richter, Ryan E Tyler, Ethan J Greenblatt, J Wade Harper, Ron R Kopito
ZUSAMMENFASSUNG
Proteins that fail to correctly fold or assemble into oligomeric complexes in the endoplasmic reticulum (ER) are degraded by a ubiquitin- and proteasome-dependent process known as ER-associated degradation (ERAD). Although many individual components of the ERAD system have been identified, how these proteins are organized into a functional network that coordinates recognition, ubiquitylation and dislocation of substrates across the ER membrane is not well understood. We have investigated the functional organization of the mammalian ERAD system using a systems-level strategy that integrates proteomics, functional genomics and the transcriptional response to ER stress. This analysis supports an adaptive organization for the mammalian ERAD machinery and reveals a number of metazoan-specific genes not previously linked to ERAD.