Merck
  • Preparation and optimization of chlorophene-loaded nanospheres as controlled release antimicrobial delivery systems.
Preparation and optimization of chlorophene-loaded nanospheres as controlled release antimicrobial delivery systems.
Pharmaceutical development and technology (2014-09-16)
Hathaichanok Phuengkham, Veerawat Teeranachaideekul, Malyn Chulasiri, Norased Nasongkla
ZUSAMMENFASSUNG
Chlorophene-loaded nanospheres with various formulation parameters were evaluated. The optimal formulation was found at 0.1% w/v of poloxamer 407, 15 mL of ethyl acetate and 20% initial chlorophene loading that provided the suitable size (179 nm), the highest loading content (19.2%), encapsulation efficiency (88.0%) and yield (91.6%). Moreover, encapsulation of chlorophene in nanospheres was able to prolong and sustain drug release over one month. Chlorophene-loaded nanospheres were effective against Staphylococcus aureus (S. aureus) and Candida albicans (C. albicans), the main cause of hospital-acquired infections. Chlorophene-loaded nanospheres were effective against S. aureus (>46 µg/mL) and C. albicans (>184 µg/mL). These nanospheres appeared to have profound effect on the time-dependent hemolytic activity due to gradual release of chlorophene. At the concentration of 46 µg/mL, nearly no HRBC hemolysis in 24 h compared to 80% of hemolysis from free drug. In conclusion, polymeric nanospheres were successfully fabricated to encapsulate chlorophene which can eliminate inherent toxicity of drugs and have potential uses in prolonged release of antimicrobial.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
2-Benzyl-4-chlorphenol, 95%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.