Merck
  • Efficient pDNA Delivery Using Cationic 2-Hydroxypropyl-β-Cyclodextrin Pluronic-Based Polyrotaxanes.
Efficient pDNA Delivery Using Cationic 2-Hydroxypropyl-β-Cyclodextrin Pluronic-Based Polyrotaxanes.
Macromolecular bioscience (2015-08-11)
Vivek Badwaik, Yawo Mondjinou, Aditya Kulkarni, Linjia Liu, Asher Demoret, David H Thompson
ZUSAMMENFASSUNG
A family of cationic Pluronic-based polyrotaxanes (PR(+)), threaded with 2-hydroxypropyl-β-cyclodextrin (HPCD), was synthesized for pDNA delivery into multiple cell lines. All PR(+) formed highly stable, positively charged pDNA complexes that were < 250 nm in diameter. The cellular uptake and pDNA transfection efficiencies of the PR(+):pDNA complexes was enhanced relative to the commercial transfection standards L2K and bPEI, while displaying similar or lower toxicity profiles. Charge density and threading efficiency of the PR(+) agent significantly influenced the colloidal stability and physical properties of the complexes, which impacted their intracellular transfection efficiencies. Taken together, our results suggest that HPCD: Pluronic PR(+) can be used as potent vectors for pDNA-based therapeutics.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Cholesterinchlorameisensäureester, 95%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.