- alpha-Synuclein regulation of the dopaminergic transporter: a possible role in the pathogenesis of Parkinson's disease.
alpha-Synuclein regulation of the dopaminergic transporter: a possible role in the pathogenesis of Parkinson's disease.
FEBS letters (2004-05-12)
Anita Sidhu, Christophe Wersinger, Philippe Vernier
PMID15135042
ZUSAMMENFASSUNG
Parkinson's disease (PD) is a slow progressive neurodegenerative disorder. Recent evidence suggests a central role for alpha-synuclein, a protein of unknown function, in the genesis of PD. The phenomenon of selective degeneration of dopaminergic neurons in PD may be linked to the potential toxicity of dopamine itself and aberrations in the processes which regulate dopamine content may underlie the pathogenesis of this disease. Here, we review a vital role of alpha-synuclein in the modulation of dopamine transporter (DAT) function, and describe how disruption of this modulatory process permits increased re-uptake of high levels of intracellular dopamine by DAT, causing profound neurotoxicity.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
α-Synuklein human, recombinant, expressed in E. coli, N-terminal histidine tagged, ≥90% (SDS-PAGE), lyophilized powder
Preise und Verfügbarkeit sind derzeit nicht verfügbar.