Merck
Sequential domain assembly of ribosomal protein S3 drives 40S subunit maturation.
Nature communications (2016-02-03)
Valentin Mitterer, Guillaume Murat, Stéphane Réty, Magali Blaud, Lila Delbos, Tamsyn Stanborough, Helmut Bergler, Nicolas Leulliot, Dieter Kressler, Brigitte Pertschy
ZUSAMMENFASSUNG
Eukaryotic ribosomes assemble by association of ribosomal RNA with ribosomal proteins into nuclear precursor particles, which undergo a complex maturation pathway coordinated by non-ribosomal assembly factors. Here, we provide functional insights into how successive structural re-arrangements in ribosomal protein S3 promote maturation of the 40S ribosomal subunit. We show that S3 dimerizes and is imported into the nucleus with its N-domain in a rotated conformation and associated with the chaperone Yar1. Initial assembly of S3 with 40S precursors occurs via its C-domain, while the N-domain protrudes from the 40S surface. Yar1 is replaced by the assembly factor Ltv1, thereby fixing the S3 N-domain in the rotated orientation and preventing its 40S association. Finally, Ltv1 release, triggered by phosphorylation, and flipping of the S3 N-domain into its final position results in the stable integration of S3. Such a stepwise assembly may represent a new paradigm for the incorporation of ribosomal proteins.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonale ANTI-FLAG® M2-Peroxidase (HRP) in Maus hergestellte Antikörper, clone M2, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-Kaninchen-IgG-(Gesamtmolekül)-Peroxidase in Ziege hergestellte Antikörper, affinity isolated antibody
Sigma-Aldrich
Monoklonaler Anti-Polyhistidin−Peroxidase-Antikörper der Maus in Maus hergestellte Antikörper, clone HIS-1, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Calmodulin-Bindungsprotein-Epitop-Tag-Antikörper, Upstate®, from rabbit
Sigma-Aldrich
Anti-RPS5 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution