Merck
  • Silencing of PTK7 in colon cancer cells: caspase-10-dependent apoptosis via mitochondrial pathway.
Silencing of PTK7 in colon cancer cells: caspase-10-dependent apoptosis via mitochondrial pathway.
PloS one (2010-11-26)
Ling Meng, Kwame Sefah, Meghan B O'Donoghue, Guizhi Zhu, Dihua Shangguan, Afshan Noorali, Yan Chen, Lei Zhou, Weihong Tan
ZUSAMMENFASSUNG
Protein tyrosine kinase-7 (PTK7) is a catalytically inactive receptor tyrosine kinase (RTK). PTK7 is upregulated in many common human cancers, including colon cancer, lung cancer, gastric cancer and acute myeloid leukemia. The reason for this up-regulation is not yet known. To explore the functional role of PTK7, the expression of PTK7 in HCT 116 cells was examined using small interference (siRNA)-mediated gene silencing. Following transfection, the siRNA successfully suppressed PTK7 mRNA and protein expression. Knocking down of PTK7 in HCT 116 cells inhibited cell proliferation compared to control groups and induced apoptosis. Furthermore, this apoptosis was characterized by decreased mitochondrial membrane potential and activation of caspase-9 and -10. Addition of a caspase-10 inhibitor totally blocked this apoptosis, suggesting that caspase-10 may play a critical role in PTK7-knockdown-induced apoptosis, downstream of mitochondria. These observations may indicate a role for PTK7 in cell proliferation and cell apoptosis and may provide a potential therapeutic pathway for the treatment of a variety of cancers.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Z-Val-Asp(O-Me)-Val-Ala-Asp(O-Me) fluoromethyl ketone, powder, ≥95%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Ptk7
Preise und Verfügbarkeit sind derzeit nicht verfügbar.