Merck
Mechanism of action of the multikinase inhibitor Foretinib.
Cell cycle (Georgetown, Tex.) (2011-11-22)
Maeva Dufies, Arnaud Jacquel, Guillaume Robert, Thomas Cluzeau, Alexandre Puissant, Nina Fenouille, Laurence Legros, Sophie Raynaud, Jill-Patrice Cassuto, Fréderic Luciano, Patrick Auberger
ZUSAMMENFASSUNG
Mitotic catastrophe (MC) is induced when stressed cells enter prematurely or inappropriately into mitosis and can be caused by ionizing radiation and anticancer drugs. Foretinib is a multikinase inhibitor whose mechanism of action is incompletely understood. We investigated here the effect of Foretinib on chronic myelogenous leukemia (CML) cell lines either sensitive (IM-S) or resistant (IM-R) to the tyrosine kinase inhibitor Imatinib. Foretinib decreased viability and clonogenic potential of IM-S and IM-R CML cells as well. Foretinib-treated cells exhibited increased size, spindle assembly checkpoint anomalies and enhanced ploidy that collectively evoked mitotic catastrophe (MC). Accordingly, Foretinib-stimulated CML cells displayed decreased expression of Cdk1, Cyclin B1 and Plk1. In addition, Foretinib triggered caspase-2 activation that precedes mitochondrial membrane permeabilization. Accordingly, z-VAD-fmk and a caspase-2 siRNA abolished Foretinib-mediated cell death but failed to affect MC, indicating that Foretinib-mediated apoptosis and MC are two independent events. Anisomycin, a JNK activator, impaired Foretinib-induced MC and inhibition or knockdown of JNK phenotyped its effect on MC. Moreover, we found that Foretinib acted as a potent inhibitor of JNK. Importantly, Foretinib exhibited no or very little effect on normal peripheral blood mononuclear cells, monocytes or melanocytes cells but efficiently inhibited the clonogenic potential of CD34+ cell from CML patients. Collectively, our data show that the multikinase inhibitor Foretinib induces MC in CML cells and other cell lines via JNK-dependent inhibition of Plk1 expression and triggered apoptosis by a caspase 2-mediated mechanism. This unusual mechanism of action may have important implications for the treatment of cancer.