Merck
  • Protein kinase A inhibits leukotriene synthesis by phosphorylation of 5-lipoxygenase on serine 523.
Protein kinase A inhibits leukotriene synthesis by phosphorylation of 5-lipoxygenase on serine 523.
The Journal of biological chemistry (2004-07-29)
Ming Luo, Sandra M Jones, Susan M Phare, Michael J Coffey, Marc Peters-Golden, Thomas G Brock
ZUSAMMENFASSUNG
Leukotrienes (LTs) are lipid messengers generated by leukocytes that drive inflammation and modulate neighboring cell function. The synthesis of LTs from arachidonic acid is initiated by the enzyme 5-lipoxygenase (5-LO). We report for the first time that LT synthesis is inhibited by the direct action of protein kinase A (PKA) on 5-LO. The catalytic subunit of PKA directly phosphorylated 5-LO in vivo and in vitro and inhibited activity in intact cells and in vitro. Mutation of Ser-523 on human 5-LO prevented phosphorylation by PKA and restored LT synthesis. Treatment with PKA activators inhibited LTB(4) synthesis in 3T3 cells expressing wild type 5-LO but not in cells expressing the S523A mutant of 5-LO. The mechanism of inhibition of LTB(4) synthesis did not involve either reduced membrane association of activated 5-LO or redistribution of 5-LO from the nucleus to the cytoplasm. Instead, PKA phosphorylation of recombinant 5-LO inhibited in vitro activity, as did co-transfection of cells with 5-LO plus the catalytic subunit of PKA. Also, substitution of Ser-523 with glutamic acid, mimicking phosphorylation, resulted in the total loss of 5-LO activity. These results indicate that PKA phosphorylates 5-LO on Ser-523, which inhibits the catalytic activity of 5-LO and reduces cellular LT generation. Thus, PKA activation, as can occur in response to adenosine, prostaglandin E(2), beta-adrenergic agonists, and other mediators, is a means of directly reducing 5-LO activity and LT synthesis that may be important in limiting inflammation and maintaining homeostasis.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Proteinkinase Katalytische A-Untereinheit aus Rinderherz, ≥9 units/μg protein (cyclic-AMP is not required for this activity), lyophilized (white powder to sticky mass to hard pellet)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.